11666703001
Roche
Buffers in a Box, Premixed TBE Buffer, 10x
solution, clear colorless, pkg of 4 L
Synonym(s):
TBE buffer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solution
Quality Level
packaging
pkg of 4 L
manufacturer/tradename
Roche
color
clear colorless
pH
~8.4
storage temp.
20-25°C
General description
Buffers in a Box, Premixed TBE Buffer, 10x is a stable 10x solution of Tris-borate-EDTA (TBE), filtered through 0.2 μm pore size membrane, resistant to enzymatic and nonenzymatic degradation. Each lot is tested for the absence of nuclease or protease activity, and stores easily at room temperature.
Application
Buffers in a Box, Premixed TBE Buffer, 10x has been used to prepare denaturing 9.3 % polyacrylamide gel for electrophoresis.
Quality
Absence of contaminants: free of DNase and RNase according to the current Quality Control procedures.
Physical form
Solution, clear, colorless
Other Notes
For life science research only. Not for use in diagnostic procedures.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Methods in molecular biology (Clifton, N.J.), 1125, 187-208 (2014-03-05)
Genome-wide analysis of gene expression has changed the RNA world. Recent techniques leading to this revolution have been the use of cross-linking and immunoprecipitation (CLIP) combined with high-throughput sequencing (HITS-CLIP) to determine sites on nascent mRNAs to which RNA-binding proteins
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service