Skip to Content
Merck
All Photos(2)

Key Documents

11427598910

Roche

Biotin-16-ddUTP

≥98% (HPLC), solution, 1 mM, suitable for hybridization

Synonym(s):

Bio-16-ddUTP tetralithium salt, 5-(N-[N-Biotinyl-ε-aminocaproyl-γ-aminobutyryl]-3-aminoallyl)-2′,3′-dideoxyuridine 5′-triphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H52N7O17P3S
CAS Number:
Molecular Weight:
931.78
MDL number:
UNSPSC Code:
41116100

Quality Level

Assay

≥98% (HPLC)

form

solution

mol wt

955.5 (biotin-16-ddUTP-Li4)

packaging

pkg of 25 μL (25 nmol; 1mM)

manufacturer/tradename

Roche

concentration

1 mM

technique(s)

hybridization: suitable

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Biotin-16-ddUTP, tetralithium salt; 1mM

Application

  • Biotin-16-ddUTP is used as a substrate for: Terminal Transferase
  • DNA polymerase I (Holoenzyme and Klenow fragment)
  • T4 DNA polymerase
  • Taq DNA polymerase and reverse transcriptase (e.g.,Transcriptor).
Biotin-16-ddUTP is preferentially used for 3′-end labeling of oligonucleotides with Terminal Transferase, recombinant and labeling DNA.
  • The biotin-labeled oligonucleotide can be used as a hybridization probe for: DNA and RNA transfers
  • Colony and plaque screening
  • In situ hybridization
In addition to common hybridization techniques, biotin-labeled oligonucleotides are specifically useful for screening expression libraries for sequence-specific DNA-binding proteins, for example, transcription factors.
The labeled oligomer can be subsequently detected by ELISA using the Streptavidin-AP conjugate for nucleic acid detection.
Oligonucleotides are enzymatically labeled at their 3′ end with Terminal Transferase by incorporation of a single biotin-labeled dideoxyuridine- triphosphate (biotin-ddUTP).
It has been used in the biotinylation of in vitro transcribed tmRNA during surface plasmon resonance (SPR) technique. It has been used in the standard nick translation reaction for tumor DNA labelling during comparative genomic hybridization.

Features and Benefits

Biotin is bound to dideoxyuridine triphosphate via an amide linkage.

Quality

Typical analysis: At least 85% Biotin-16-ddUTP (HPLC, area%).

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Microarray-based method for detection of unknown genetic modifications.
Torstein T, et al.
BMC biotechnology, 7(1), 91-91 (2007)
Peter Liebisch et al.
British journal of haematology, 122(2), 193-201 (2003-07-09)
Chromosomal abnormalities, such as 13q deletions, are emerging as important prognostic factors in multiple myeloma. Fluorescence in situ hybridization (FISH) using specific DNA probes is the technique most widely used for the determination of genomic aberrations in this disease. The
Highly polymorphic microsatellite markers for the short-snouted seahorse (Hippocampus hippocampus), including markers from a closely related species the long-snouted seahorse (Hippocampus guttulatus).
van de Vliet M S, et al.
Conservation genetics resources, 1(1), 93-93 (2009)
Takahiro Okada et al.
Bioscience, biotechnology, and biochemistry, 68(11), 2319-2325 (2004-11-27)
Escherichia coli ribosomal protein S1 is composed of six repeating homologous oligonucleotide/oligosaccharide-binding fold (OB folds). In trans-translation, S1 plays a role in delivering transfer-messenger RNA (tmRNA) to stalled ribosomes. The second OB fold of S1 was found to be protected
Hadar Golan Berman et al.
International journal of molecular sciences, 22(11) (2021-06-03)
Cisplatin is a chemotherapy drug that kills cancer cells by damaging their DNA. In human cells, this damage is repaired primarily by nucleotide excision repair. While cisplatin is generally effective, many cancers exhibit initial or acquired resistance to it. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service