Skip to Content
Merck
All Photos(3)

Key Documents

11202375001

Roche

DOTAP Liposomal Transfection Reagent

>99% (TLC), liquid, suitable for transfection

Synonym(s):

DOTAP methosulfate, liposomal transfection reagent, dotap, transfection reagent, dotap, N-(2,3-Dioleoyloxy-1-propyl)trimethylammonium methyl sulfate, Transfection reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H83NO8S
CAS Number:
Molecular Weight:
774.19
MDL number:
UNSPSC Code:
41106502
PubChem Substance ID:

description

N-[1-(2,3-Dioleoyloxy)propyl]-N,N,N-trimethylammonium methyl-sulfate

Quality Level

Assay

>99% (TLC)

form

liquid

packaging

pkg of 5 × 400 μL (10 U/μl)

manufacturer/tradename

Roche

technique(s)

transfection: suitable

storage temp.

2-8°C

SMILES string

[H]C(C[N+](C)(C)C)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.O=S([O-])(OC)=O

InChI

1S/C42H80NO4.CH4O4S/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-41(44)46-39-40(38-43(3,4)5)47-42(45)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2;1-5-6(2,3)4/h20-23,40H,6-19,24-39H2,1-5H3;1H3,(H,2,3,4)/q+1;/p-1/b22-20-,23-21-;

InChI key

RSMRWWHFJMENJH-LQDDAWAPSA-M

Looking for similar products? Visit Product Comparison Guide

General description

DOTAP Liposomal Transfection Reagent is a liposome formulation of the cationic lipid DOTAP. Unlike other liposomal reagents, it produces highly efficient transfections both in the presence and absence of serum. It can also be successfully used for in vivo applications. Mixing DOTAP with DNA results in spontaneously formed stable complexes. These complexes can be added directly to the cell culture medium. This method of DNA transfer is very gentle, avoiding the cytotoxic effects commonly encountered with lipofection or other transfection methods.

Application

Cationic liposome-forming compound for transfection of DNA, RNA and other negatively charged molecules into eukaryotic cells. Mixing DOTAP with DNA results in spontaneously formed stable complexes that can be directly added to cell culture medium. These complexes fuse with the cell membrane and release DNA into the cytoplasm. The cells are transfected efficiently without cytotoxic effects. Use approximately 5-10 μg DOTAP per μg DNA and approximately 10-20 μg DOTAP per mL cell culture medium. The optimal working concentration and transfection time depends in part on the cell line used, and the type of material (RNA, DNA, protein) loaded.
The DOTAP (N-[1-(2,3-Dioleoyloxy)propyl]-N,N,N-trimethylammonium methyl-sulfate) Liposomal Transfection Reagent is a liposome formulation of the cationic lipid DOTAP. DOTAP can be used for the highly efficient transfection of DNA including yeast artificial chromosomes (YACs) into eukaryotic cells for transient or stable gene expression, and is also suitable for the efficient transfer of other negatively charged molecules, such as RNA, oligonucleotides, nucleotides, ribonucleo-protein (RNP) complexes, and proteins into research samples of mammalian cells.

Features and Benefits

  • Easy to use: The lipid dispersion is simply mixed with the DNA solution, then applied directly to the cells
  • Highly effective: 5 to 100-fold more effective than the calcium phosphate or DEAE-dextran method
  • Gentle: No cytotoxic effects
  • Flexible: Equally effective in the presence or absence of serum. Effective on a wide range of species (e.g., insect cells, mammalian cells) and a variety of different cell types. Can be used for either transient or stable transfection procedures.
  • Aqueous dispersion (liposomes) in MBS (MES-buffered saline, pH 6.2) (bottled under argon), 1mg/ml, sterile-filtered.

Preparation Note

Working concentration: Approximately 5 - 10μg DOTAP/μg DNA, and 10 - 20μg DOTAP/ml cell culture medium.
The optimal working concentration is dependent on several parameters, including the cell line being used, concentration and type of nucleic acid (DNA, RNA), and incubation time. It is important to optimize the transfection conditions for the individual cell type studied.
Cytotoxicity: Not cytotoxic up to a concentration of 100μg/ml (PBLs, HeLa cells).

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Rodríguez et al.
International journal of cancer, 81(5), 785-792 (1999-05-18)
The effect of incubations with anti-sense phosphorothioate oligonucleotides directed toward sequences of dihydrofolate reductase (DHFR) RNA has been tested on Chinese hamster ovary cells. The selected targets were the 5'-untranslated region, the translational start, the splice sites and branch point
S Marshak et al.
Diabetes, 48(6), 1230-1236 (1999-05-26)
In type 2 diabetes, chronic hyperglycemia has been suggested to be detrimental to beta-cell function, causing reduced glucose-stimulated insulin secretion and disproportionately elevated proinsulin. In the present study, we investigated the effect on several beta-cell functions of prolonged in vitro
D Chakravarti et al.
Cell, 96(3), 393-403 (1999-02-20)
Nucleosomal histone modification is believed to be a critical step in the activation of RNA polymerase II-dependent transcription. p300/CBP and PCAF histone acetyltransferases (HATs) are coactivators for several transcription factors, including nuclear hormone receptors, p53, and Stat1alpha, and participate in
S M Richards et al.
Journal of cell science, 112 ( Pt 10), 1519-1528 (1999-04-23)
The anion exchangers (AE) are encoded by a multigenic family that comprises at least three genes, AE1, AE2 and AE3, and numerous splicoforms. Besides regulating intracellular pH (pHi) via the Cl-/HCO3- exchange, the AEs exert various cellular functions including generation
B Weiss et al.
Cellular and molecular life sciences : CMLS, 55(3), 334-358 (1999-05-06)
Agents that produce their effects through an antisense mechanism offer the possibility of developing highly specific alternatives to traditional pharmacological antagonists, thereby providing a novel class of therapeutic agents, ones which act at the level of gene expression. Among the

Articles

This brief webinar provides an overview of what transfection is and the methods that are used to introduce DNA or RNA into eukaryotic cells.

Related Content

Browse our convenient transfection reagent selection guide to match the best reagent for your specific cell line and application needs.

Browse our convenient transfection reagent selection guide to match the best reagent for your specific cell line and application needs.

Browse our convenient transfection reagent selection guide to match the best reagent for your specific cell line and application needs.

Browse our convenient transfection reagent selection guide to match the best reagent for your specific cell line and application needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service