Skip to Content
Merck

VMWP04700

MCE Membrane Filter, 0.05 μm Pore Size

greener alternative

MF-Millipore, filter diam. 47 mm, hydrophilic

Synonym(s):

MF-Millipore Membrane Filter, 0.05 µm pore size

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
0.05 μm pore size, 72 % porosity
Material:
mixed cellulose esters (MCE) membrane, plain filter, white filter
Feature:
hydrophilic
Diam.:
47 mm
Sterility:
non-sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

description

47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore, Millipore

sustainability

Greener Alternative Product

parameter

0.25 L/min-cm2 air flow rate, 0.74 mL/min-cm2 water flow rate, 75 °C max. temp.

diam.

47 mm

filter diam.

47 mm

thickness

100 μm

gravimetric extractables

1.5%

refractive index

n/D 1.5

matrix

MF-Millipore

pore size

0.05 μm pore size, 72 % porosity

bubble point

≥17.6 bar, air with water at 23 °C

greener alternative category

shipped in

ambient

General description

MF-Millipore membrane comprises cellulose acetate and cellulose nitrate. These biologically inert mixtures have wide analytical and research applications.
Need a custom membrane filter? Click here to request a custom membrane filter or unique and large package sizes that may not be available via our website.
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

Microdialysis of DNA and proteins

Features and Benefits

  • Applicable for biological and environmental monitoring applications.
  • Autoclavable and Compatible with ethylene oxide and gamma irradiation sterilization methods.

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
  • Storage Statement: Store in dry location away from heat source
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


signalword

Danger

Storage Class

4.1B - Flammable solid hazardous materials

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Irrit. 2 - Flam. Sol. 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Janet R Donaldson et al.
The Journal of biological chemistry, 281(39), 28811-28821 (2006-08-10)
RuvABC is a complex that promotes branch migration and resolution of Holliday junctions. Although ruv mutants are hypersensitive to UV irradiation, the molecular event(s) that necessitate RuvABC processing in vivo are not known. Here, we used a combination of two-dimensional
Fate and immunotoxic effects of silver nanoparticles on rainbow trout in natural waters
Bruneau A, et al.
Journal of nanomedicine & nanotechnology, 6(03), 290-290 (2015)



Global Trade Item Number

SKUGTIN
VMWP0470004053252467981