Select a Size
About This Item
Quality Level
material
mixed cellulose esters (MCE) membrane, plain filter, white filter
description
13 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs
sterility
non-sterile
feature
hydrophilic
manufacturer/tradename
MF-Millipore™, Millipore
sustainability
Greener Alternative Product
parameter
0.25 L/min-cm2 air flow rate, 0.74 mL/min-cm2 water flow rate, 75 °C max. temp.
diam.
13 mm
filter diam.
13 mm
thickness
100 μm
gravimetric extractables
1.5%
refractive index
n/D 1.5
matrix
MF-Millipore™
pore size
0.05 μm pore size, 72 % porosity
bubble point
≥17.6 bar, air with water at 23 °C
greener alternative category
shipped in
ambient
General description
MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.
Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Application
Other Notes
- Organism Retention: Microorganism
- Mode of Action: Filtration (size exclusion)
- Application: General laboratory filtration
- Intended Use: Retention or removal of biological contaminants
- Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
- Storage Statement: Store in dry location away from heat source
- Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.
Legal Information
Still not finding the right product?
Explore all of our products under MCE Membrane Filter, 0.05 μm Pore Size
signalword
Danger
Storage Class
4.1B - Flammable solid hazardous materials
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Irrit. 2 - Flam. Sol. 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| VMWP01300 | 04053252359125 |


