Skip to Content

TMRES

mPAGE® TurboMix Resolving Solution 216ML

Premixed resolving gel solution for casting bis-tris polyacrylamide gels

Synonym(s):

Bis-Tris gel resolving solution, resolving gel solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NC.01
UNSPSC Code:
41105319
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

0.2 μm filtered

form

liquid

feed ratio

29:1

storage condition

protect from light

storage temp.

2-8°C

General description

mPAGE® TurboMix Resolving Solution is a 20% acrylamide buffer solution used to make bis-tris polyacrylamide gels. It can be diluted with DI water to make any gel percentage between 8% and 15%. After dilution, add APS and TEMED and then pour gels to the desired height.

Application

Premixed resolving gel solution for casting bis-tris polyacrylamide gels.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


signalword

Danger

target_organs

Peripheral nervous system

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Health hazardExclamation mark

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library