Skip to Content
Merck

SCT040

BioTracker Green Free H2O2Dye

Fluorescent probe for detection of extracellular hydrogen peroxide (H2O2) and free H2O2 in solutions.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized

application(s)

cell analysis

detection method

fluorometric

General description

Hydrogen peroxide (H2O2) is involved in therapeutic processes such as wound healing, anti-bacterial defense, stem cell proliferation, and an adaptive response in astrocytes that leads to neuronal protection. However, over-production of H2O2 exerts toxic effects on the cell and its surrounding environment. Over-production of H2O2 is connected to serious pathological conditions such as cancer, ageing, diabetes, and neurodegenerative diseases.BioTracker Green free H2O2 dye is a fluorescent probe that fluoresces upon reaction with H2O2 , but does not react with other ROS such as hydroxyradical (-OH), superoxide (O2-), hypochlorous acid (HOCl), singlet oxygen (1O2), and nitric oxide (NO). This dye cannot penetrate cell membrane but can be used for detection of extracellular H2O2 and in free solutions. Spectral Properties:Absorbance (max): 492 nmEmission (max): 516 nm (Green)Research Categories:AgingNeurodegenerative Diseases

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library