Skip to Content
Merck

Skip To

S2GPU02RE

Millipore® Stericup® Quick Release Vacuum Filtration System

pore size 0.22 μm, polyethersulfone membrane, filter capacity 250 mL, pack of 12 ea

Millipore® Stericup® Quick Release Vacuum Filtration System
1 of 3 reviewers received a sample product or took part in a promotion

Synonym(s):

SCGPU02RE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41104914
NACRES:
NB.24
eCl@ss:
32031623
Pore size:
0.22 μm pore size
Filter capacity:
250 mL
Packaging:
pack of 12 ea
Material:
polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing
Sterility:
sterile
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

sterility

sterile

product line

Stericup®

feature

hydrophilic

packaging

pack of 12 ea

parameter

45 °C max. temp.

description

2.4 mL (hold-up volume)

technique(s)

sterile filtration: suitable

H

164 mm

diam.

12.5 cm

filter capacity

250 mL

filter diam.

73 mm

filtration area

40 cm2

matrix

Express PLUS

pore size

0.22 μm pore size

fitting

inlet funnel, outlet flask (Receiver)

shipped in

ambient

storage temp.

no temp limit

General description

This item is a direct replacement for SCGPU02RE
Work with ease and filter with confidence with Stericup® and Steritop® Filtration Systems.

Stericup® and Steritop® sterile filtration devices use reliable, high-performance membranes for optimum sterile filtration of laboratory reagents, buffers, and media. These devices combine superior flow rates and throughput with low non-specific binding and a stable, no-tip design.

This Stericup® 0.22 μm pore size polyethersulfone (PES) membrane filter unit combines a 250 mL bottle-top with a 250 mL receiver flask and cap for processing and storage.

Application

For Research Use Only
Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.
  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions

Features and Benefits

  • The Quick Release filter funnel disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
  • Bold product identification on the Steritop® filter funnel plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
  • Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
  • Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
  • Click seal cap rests on the side to avoid risk of contamination.
  • Receiver bottles are stackable to save storage space.

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S2GPU01RES2GPU11RES2GPU10RE
pore size

0.22 μm pore size

pore size

0.22 μm pore size

pore size

0.22 μm pore size

pore size

0.22 μm pore size

filter capacity

250 mL

filter capacity

150 mL

filter capacity

1000 mL

filter capacity

500 mL

sterility

sterile

sterility

-

sterility

sterile

sterility

sterile

material

polyethersulfone membrane, polystyrene funnel(s), polystyrene bottle, polystyrene housing

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

packaging

pack of 12 ea

packaging

pack of 12 ea

packaging

pack of 12 ea

packaging

pack of 12 ea

fitting

inlet funnel, outlet flask (Receiver)

fitting

inlet funnel, outlet flask (Receiver)

fitting

inlet funnel, outlet flask (Receiver)

fitting

inlet funnel, outlet flask (Receiver)


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–10 of 16 Questions  
  1. Buongiorno. I filtri resistono ad inchiostri UV a base acrilica, all'acetone e all'alcool isopropilico?

    1 answer
    1. This product is composed of polyethersulfone and polystyrene materials. Hence, it is not recommended for use with acetone.
      Please see the chemical compatibility chart: https://www.sigmaaldrich.com/technical-documents/technical-article/analytical-chemistry/filtration/chemical-compatibility-of-filter-components?utm_source=redirect&utm_medium=promotional&utm_campaign=FilterChemicalCompatibility

      However, the product's compatibility with acrylic-based UV inks and isopropyl alcohol has not been validated.
      Please see the user guide for more information:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/337/924/20199204w-ms.pdf

      Helpful?

  2. What leachables are detected during filtration processes using Stericup® filters?

    1 answer
    1. We have not performed extractables/leachables studies for our Stericup® filters.

      Helpful?

  3. Can Stericup® bottles be placed inside freezers for long-term storage of media?

    1 answer
    1. You can successfully freeze and store many aqueous solutions (such as culture media) in Stericup® bottles at temperatures up to -20 °C (-4 °F). It is strongly recommended that you run a sample stability trial under your actual storage conditions prior to using Stericup® bottles for frozen storage.

      Helpful?

  4. The User Guide says “Use only glass or plastic bottles designed for vacuum applications”. Can you specify which bottles are compatible?

    1 answer
    1. Steritop® Quick Release bottle top units require a vacuum-rated receiver bottle with either a GL33 or GL45 mm closure thread as specified. Steritop® E bottle top units require a vacuum-rated receiver bottle with a GL45 mm closure thread. Bottles no larger than 2 L should be used.
      Glass bottles, GL33:
      • Wheaton media bottles (glass), 500 mL (Product Z364886)
      • Wheaton media bottles (glass), 1 L (Product Z364894)
      Glass bottles, GL45
      • Corning® glass media storage bottle, 125 mL (Product CLS1399125)
      • Corning® glass media storage bottle, 250 mL (Product CLS1399250)
      • Corning® glass media storage bottle, 500 mL (Product CLS1399500)
      • Corning® glass media storage bottle, 1 L (Product CLS13991L)
      • Pyrex® round media storage bottle, 500 mL (Product CLS1395500)
      • Pyrex® round media storage bottle, 1 L (Product CLS13951L)
      • KIMBLE® KIMAX® Media bottle, 250 mL (Product DWK14395250)
      • KIMBLE® KIMAX® Media bottle, 500 mL (Product DWK14395500)
      • KIMBLE® KIMAX® Media bottle, 1 L (Product DWK143951000)
      Plastic (pre-sterile) bottles, GL45:
      • Stericup® Quick Release Receiver flask, 250 mL (Product S200B02RE)
      • Stericup® Quick Release Receiver flask, 500 mL (Product S200B05RE)
      • Stericup® Quick Release Receiver flask, 1000 mL (Product S200B10RE)

      Helpful?

  5. What tubing can be used with Stericup® and Steritop® vacuum filters?

    What tubing can be used with Stericup® and Steritop® vacuum filters?

    1 answer
    1. Stericup® and Steritop® filters are fitted with a 1/4” tube adapter. We recommend 3/16 in. (4.8 mm) ID silicone tubing to fit this tube adapter: https://www.sigmaaldrich.com/product/mm/xx7100004

      Helpful?

  6. Are Stericup® and Steritop® filters GMP-certified?

    1 answer
    1. No, Stericup® and Steritop® devices are not GMP-approved.

      Helpful?

  7. What is the chemical compatibility of the various Stericup® and Steritop® filters?

    What is the chemical compatibility of the various Stericup® and Steritop® filters?

    1 answer
    1. Helpful?

  8. What is the maximum vacuum pressure for filtration?

    1 answer
    1. 700 mm Hg (27.5” Hg, 0.9 bar) differential vacuum at 25 °C (77 °F)

      Helpful?

  9. Can we filter oil-type liquids with Stericup® filters?

    Can we filter oil-type liquids with Stericup® filters?

    1 answer
    1. Likely not, due to pore size/possible fouling/chemical compatibility. Refer to our guide, "Chemical compatibility of filter components": https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/302/855/membrane-chemical-compatibility-ds2532en-ms.pdf

      Helpful?

  10. What is the maximum processing volume for Stericup® and Steritop® filters? Can I add more volume after initial filtration?

    1 answer
    1. All Stericup® and Steritop® filters were developed for single-use application. The maximum volume is dependent on the receiver bottle capacity. Stericup® filters come with a receiver bottle with a specified maximum volume. For Steritop® filters, the maximum recommended volume is 2 L with an appropriate-sized receiver bottle. Larger processing volumes can result in membrane fouling, increase chance of spillage and contamination, and lead to possible airlock*.
      *Note that devices with PES membranes can airlock if filtered to dryness. Some liquid must be left on top of the filter, then the system stopped, receiver changed or emptied, and more sample volume added before restart. This increases the chance of spillage.

      Helpful?

1–10 of 16 Questions  

Reviews

Active Filters

  1. Columbia, MO
    • Reviews 4
    • Votes 0
    5 out of 5 stars.

    Easy to use filter units that work well

    Sigma is known for its high quality of chemical and biochemical reagents, and this is no exception.

    Helpful?

  2. 2 Ratings-only reviews