Skip to Content
Merck

RAWP14250

MCE Membrane Filter, 1.2 μm Pore Size

MF-Millipore, filter diam. 142 mm, hydrophilic

Synonym(s):

MF-Millipore Membrane Filter, 1.2 µm pore size

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
1.2 μm pore size, 82 % porosity
Material:
mixed cellulose esters (MCE) membrane, plain filter, white filter
Feature:
hydrophilic
Diam.:
142 mm
Sterility:
non-sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

description

142 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 50 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore, Millipore

parameter

20 L/min-cm2 air flow rate, 270 mL/min-cm2 water flow rate, 75 °C max. temp.

diam.

142 mm

filter diam.

142 mm

thickness

150 μm

gravimetric extractables

5%

refractive index

n/D 1.512

matrix

MF-Millipore

pore size

1.2 μm pore size, 82 % porosity

bubble point

≥0.76 bar, air with water at 23 °C

shipped in

ambient

General description

  • MF-Millipore® mixed cellulose esters membrane filter is ideal for biological and environmental monitoring applications.
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods.
  • The white filter is available with a gridded surface.

Application

Clarification of aqueous solutions

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


pictograms

Flame

signalword

Danger

Storage Class

4.1B - Flammable solid hazardous materials

Hazard Classifications

ED ENV 1 - Flam. Sol. 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library