Skip to Content
Merck

PP54

IgG, Mouse

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Technique(s):
ELISA: suitable
Application:
ELISA
Citations:
39
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

secondary antibodies

clone

polyclonal

assay

95% (SDS-PAGE)

manufacturer/tradename

Chemicon®

concentration

25 mg/mL

technique(s)

ELISA: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

Mouse IgG, whole molecule, purified from normal mouse serum. Purity: 95% by SDS-PAGE

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Physical form

Format: Purified
Protein A purified
Purified mouse polyclonal IgG in buffer containing 0.01M PBS, 0.15M sodium chloride, 0.1% sodium azide, pH 7.1.

Preparation Note

Stable for 1 year at from date of receipt.

Analysis Note

ELISA: A previous lot of this antibody was used in ELISA.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neprilysin is identical to skin fibroblast elastase: its role in skin aging and UV responses.
Morisaki, N; Moriwaki, S; Sugiyama-Nakagiri, Y; Haketa, K; Takema, Y; Imokawa, G
The Journal of Biological Chemistry null
Lin28 regulates BMP4 and functions with Oct4 to affect ovarian tumor microenvironment.
Ma, W; Ma, J; Xu, J; Qiao, C; Branscum, A; Cardenas, A; Baron, AT; Schwartz, P; Maihle, NJ; Huang, Y
Cell Cycle null
Astaxanthin attenuates the UVA-induced up-regulation of matrix-metalloproteinase-1 and skin fibroblast elastase in human dermal fibroblasts.
Suganuma K, Nakajima H, Ohtsuki M, Imokawa G
Journal of Dermatological Science null
Design and activity of multifunctional fibrils using receptor-specific small peptides.
Y Ohga, F Katagiri, K Takeyama, K Hozumi, Y Kikkawa, N Nishi, M Nomizu
Biomaterials null
Daniela F Requena et al.
Journal of neurochemistry, 110(2), 581-594 (2009-05-22)
Synaptic transmission depends on the efficient loading of transmitters into synaptic vesicles by vesicular neurotransmitter transporters. The vesicular monoamine transporter-2 (VMAT2) is essential for loading monoamines into vesicles and maintaining normal neurotransmission. In an effort to understand the regulatory mechanisms

Global Trade Item Number

SKUGTIN
PP5404053252317354

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service