Skip to Content
Merck

Skip To

P3B030D10

Pellicon® 3 Cassette with Biomax® Membrane

30 kDa, D screen, 1.14 m2

Synonym(s):

Pellicon® 3 Cassette with Biomax 30 kDa Membrane, D screen, 1.14 m2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

eCl@ss:
32031623
UNSPSC Code:
23151806
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

material

TPE seal, polyethersulfone membrane, polypropylene housing, polypropylene screen, stainless steel cap (for end; polypropylene-encapsulated)

manufacturer/tradename

Pellicon®

storage condition

do not freeze

parameter

30 psi max. reverse press. exposure at 25 °C (for 24 hours), 4-6 L/min-m2 recirculation rate, 4-8 L/min-m2 flow rate, 40 psi max. transmembrane pressure at 4-50 °C, 6.9 bar max. inlet pressure (100 psi), 80 psi max. transmembrane pressure at 4-40 °C

technique(s)

tangential flow filtration (TFF): suitable, ultrafiltration: suitable

H

4.2 cm (1.67 in.)

L

20.6 cm (8.1 in.)

W

17.8 cm (7 in.)

filtration area

1.14 m2

size

229 mL , Volume (feed), 88 mL , Volume (permeate)

gravimetric extractables

≤1500 mg/m2

matrix

Biomax®

pore size

(30 kDa)

pH

1-14

storage temp.

2-8°C

General description

Pellicon® 3 cassettes with Biomax membrane are the optimum tangential flow filtration (TFF) devices for the filtration of solutions containing therapeutical proteins, albumin, hormones, vaccines and growth factors. These advanced, high-performance cassettes are ideal for today’s higher titer therapeutic antibodies, higher viscosity feed formulations as well as the more demanding filtration processes that require higher operating pressures, temperatures and caustic cleaning regimes.From small-scale to full-scale production, Pellicon® 3 cassettes are designed for use in research, process scale-up/scale-down, applications development and full-scale manufacturing. The Pellicon® 3 design and automated manufacturing process provides unbeatable performance consistency between cassette sizes. Pellicon® 3 devices also offer greater cassette size selection for improved scale-up and scale-down process development. The streamlined design allows operators to quickly and easily handle, install and remove Pellicon® 3 cassettes. The materials of construction are compatible with a broad range of chemical cleaning.

Application

monoclonal antibodies, recombinant and non-recombinant proteins, vaccines

Features and Benefits

  • Robust, void-free membrane provides optimum productretention and performance consistency
  • Unique feed screen design enables high mass transfer, flux for higher finalconcentration
  • Fast, reliable scale up/down from lab to production scale
  • Automated manufacturing delivers unbeatable performance consistency andreliability
  • Easy to install and clean
  • Extreme temperature and chemical compatibility

Analysis Note

Gravimetric Extractables: after 24 hours in water at controlled room temperature

Other Notes

These products are manufactured in a facility which adheres to Good Manufacturing Practices.

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P3B030A10P3B030A00P3B030A05
pore size

(30 kDa)

pore size

30 kDa NMWCO

pore size

30 kDa NMWCO

pore size

30 kDa NMWCO

filtration area

1.14 m2

filtration area

-

filtration area

88 cm2

filtration area

-

technique(s)

tangential flow filtration (TFF): suitable, ultrafiltration: suitable

technique(s)

buffer exchange: suitable, protein purification: suitable (concentration)

technique(s)

buffer exchange: suitable, protein purification: suitable (concentration)

technique(s)

buffer exchange: suitable, protein purification: suitable (concentration)

matrix

Biomax®

matrix

Biomax® (Membrane)

matrix

Biomax® (Membrane)

matrix

Biomax® (Membrane)

material

TPE seal, polypropylene housing, stainless steel cap (for end; polypropylene-encapsulated), polyethersulfone membrane, polypropylene screen

material

TPE seal, polyethersulfone , polypropylene screen, stainless steel cap (for end; polypropylene-encapsulated)

material

TPE seal, polyethersulfone membrane (void free), polypropylene , polypropylene cap (for end), polypropylene screen

material

TPE seal, polyethersulfone , polypropylene screen, stainless steel cap (for end; polypropylene-encapsulated)

size

229 mL , Volume (feed), 88 mL , Volume (permeate)

size

-

size

-

size

-


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 2



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
P3B030D1004055977307078

Questions

Reviews

No rating value

Active Filters