Skip to Content
Merck

P2PLBCV01

Pellicon® 2 Mini Cassette Media: 3 kDa

Chemistry: Regenerated Cellulose (RC) Screen Type: V Device Size: 0.1 m2

Synonym(s):

Pellicon 2 Mini Ultrafiltration Module PLBC V 0.1 m2, Pellicon® 2 Mini Ultrafiltration Module

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
23151806
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Pellicon® 2 Mini Cassette Media: 3 kDa, Chemistry: Regenerated Cellulose (RC) Screen Type: V Device Size: 0.1 m2

Quality Segment

material

polypropylene screen, polyurethane adhesive, regenerated cellulose

manufacturer/tradename

Pellicon®

parameter

3.5 bar max. transmembrane pressure (50 psig) at 30 °C, 5-35 L/min recirculation rate at 0.4 bar (6 psi)

technique(s)

analytical sample preparation: suitable, protein purification: suitable

H

2.16 cm (0.85 in.)

L

21 cm (8.3 in.)

W

5.6 cm (2.2 in.)

filtration area

0.1 m2

membrane nominal area

0.1 m2

gravimetric extractables

≤1500 mg/m2

matrix

Ultracel®

pore size

3 kDa NMWCO

General description

Device Configuration: Mini Cassette
Filter Code: PLBC
Filter Type: Ultrafiltration

Features and Benefits

Type V screen (open channel)

Analysis Note

Gravimetric Extractables: after a water flush

Other Notes

Prefiltration Rquirements: 100 µm
These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library