Skip to Content
Merck

MSGVN22

Multiscreen® 96 well Plate, hydrophilic PVDF membrane

pore size 0.22 μm, non-sterile

Synonym(s):

Hydrophilic 96-Well Plate, PVDF 96-Well Plate

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
41104923
NACRES:
NB.22
eCl@ss:
32039006
Material:
flat bottom wells
Durapore PVDF membrane
clear acrylic plate
Pore size:
0.22 μm pore size
Sterility:
non-sterile
Binding type:
low binding surface
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Multiscreen® 96 well Plate, hydrophilic PVDF membrane, pore size 0.22 μm, non-sterile

material

flat bottom wells
Durapore PVDF membrane
clear acrylic plate

description

Receptor binding assays on whole cells and cell fragments, protein kinase/phosphatase precipitation assays, bead/resin based assays, solubility assay testing

sterility

non-sterile

Quality Level

usage

Automated

feature

hydrophilic
lid

packaging

pkg of 10 plates
pkg of 50 plates

manufacturer/tradename

MultiScreen®

technique(s)

activity assay: suitable (protein kinase/phosphatase precipitation)
activity assay: suitable
bead-based assay: suitable
ligand binding assay: suitable
low protein binding filtration: suitable
receptor binding assay: suitable
solubility assay: suitable

filtration area

0.28 cm2

plate size

96 well

well maximum volume

300 μL

working volume

50-250 μL

matrix

Durapore®

pore size

0.22 μm pore size

application(s)

sample preparation

binding type

low binding surface

compatibility

centrifuge & manifold

shipped in

ambient

Application

Research Category
Protein

Assays

Bead/Resin based assays

General description

Filter Code: GV
No matter what application you’re working with, Millipore has a plate developed to meet your specific needs. MultiScreen plates have a long history as a reliable tool for the advancement of drug discovery and life science research.

HTS Filter Plates (Plate catalogue numbers that start with MS)
- MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics.

- Millipore’s new MultiScreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability in both background and signal intensities specifically for biochemical screening assays.

Classic Filter Plates (Plate catalogue numbers that start with MA)
- The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.

Accessory – MultiScreenHTS Vacuum Manifold
- The MultiScreenHTS Vacuum manifold was designed specifically to work with MultiScreenHTS filter plates. The combination results in optimal filtration characteristics and seamless automation integration.

Millicell Assay Plates
- In addition to the traditional assay plate format, filter plates are also available with cell culture trays for growing and assaying suspension and adherent cell lines. Cells grown on filter plates differentiate better than those grown on traditional plastic substrates, resulting in more in vivo-like results This filter plate/tray combination can be used for a variety of cellular assays, including, transport, toxicity and screening assays.

Other Notes

Automation Compatible: Yes

Physical form

Color Code: Clear

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service