Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.24
UNSPSC Code:
41116129
Pore size:
0.4 μm pore size
Material:
plain filter, polycarbonate membrane, white filter
Feature:
easily wetted with water, hydrophilic
Sterility:
non-sterile
material
plain filter, polycarbonate membrane, white filter
sterility
non-sterile
feature
easily wetted with water, hydrophilic
manufacturer/tradename
Microfil® with Isopore
filter size
47 mm
pore size
0.4 μm pore size
input
sample type food(s)
sample type beverage(s)
sample type pharmaceutical(s)
sample(s) (environmental)
application(s)
bioburden testing
clinical
environmental
food and beverages
industrial qc
pharmaceutical
water monitoring
shipped in
ambient
General description
Isopore polycarbonate membrane filters of 0.4 μm pore size are and ready to use. These filters possess high recoveries and are more appropriate for general microbiological testing
Application
These filters find application in pharmaceuticals, Water Monitoring, Clinical Lab, Food and Beverage, Industrial QC and Environmental samples for general microbiological analysis.
Legal Information
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.