Skip to Content
Merck
All Photos(2)

Key Documents

MC0HC20CL3

Millipore

Millistak+® HC Depth Filter in Micro 20 format

greener alternative

C0HC, filtration area 20 cm², Process Development Scale

Synonym(s):

20 cm2 surface area, Female-Luer fitting connection, Millistak+® C0HC Micro 20 Depth Filter, Millistak+® HC Pod Depth Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806

Product Name

Millistak+® HC Depth Filter in Micro 20 format, C0HC, cellulose media, filtration area 20 cm2, hydrophilic

material

cellulose media
filter aid (inorganic)
glass filled polypropylene housing

Quality Level

feature

hydrophilic

manufacturer/tradename

Millistak+®

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

diam.

2.5 in. (6.4 cm)

filtration area

20 cm2

impurities

<0.25 EU/mL (LAL test)

total organic carbon (TOC) residue

720-4600 ppb

input

sample type liquid

conductivity

3.64-10.5 μS/cm

fitting

female Luer fitting

filter grade

C0HC

greener alternative category

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Device Configuration: Micro 20
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.
post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 10 L/ft2 (100 L/m2) of media surface area

Application

Perfusion bioreactor fluid

Features and Benefits

Millistak+® HC Micro 20 devices offer:

  • Improved scalability during media screening trials
  • Smaller filtration area slightly reduces feedstream volume requirements (approximately 15% less).
  • Transparent housing enables visibility of fluid flow
  • Easier to calculate flow rates based on 20 cm2 filter area


Millistak+® HC (high capacity) media offers:
  • Millistak+® HC dual-action media improves prefiltration and compresses clarification


Millistak+® Pod filters offer:
  • Low hold-up volume for greater product yield
  • Broad range of media grades offered in single and multilayer products
  • Flexible, modular format offers scalability from 5 to 12,000 liters or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces

Packaging

3 EA

Other Notes

Integrity is maintained after 2 autoclave cycles of 60 minutes at 123 °C

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

MILLISTAK+® is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn more on mAb downstream processing, more specifically clarification and the relevant associated products.

Learn more on mAb downstream processing, more specifically clarification and the relevant associated products.

Learn more on mAb downstream processing, more specifically clarification and the relevant associated products.

Learn more on mAb downstream processing, more specifically clarification and the relevant associated products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service