Skip to Content
Merck
All Photos(2)

Documents

MABT83

Sigma-Aldrich

Anti-Aggrecan Antibody, clone 6B4

clone 6B4, from mouse

Synonym(s):

Aggrecan core protein, Cartilage-specific proteoglycan core protein, CSPCP, Chondroitin sulfate proteoglycan core protein 1, Chondroitin sulfate proteoglycan 1, Aggrecan core protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6B4, monoclonal

species reactivity

bovine

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACAN(176)

General description

Aggrecan (CSPG1, CSPCP, or CSPGCP) is a chondroitin-sulfate proteoglycan belonging to the family of letican proteins. Aggrecan is a large globar structure containing multiple domains including carbohydrate-recognition domains; complement binding protein domains; immunoglobulin domains; proteoglycan tandem repeats; and in some cases, EGF-like domains. It can be degraded by aggrecanase. The N-terminus of aggrecan binds ECM hyaluronan in the presence of the linking protein, and forms complex aggregates, whereas the C-terminus binds a number of other ECM proteins including tenascins, glycolipids, and fibulins. Aggrecan is a major component of cartilage tissue and it enables this tissue to withstand compression by maintaining the viso-elastic properties of cartilage. Aggrecan may also facilitate neural maturation and plasticity by contributing to the formation of perineuronal nets in the developing CNS.

Immunogen

Ovalbumin-conjugated linear peptide corresponding to human Aggrecan.

Application

Anti-Aggrecan Antibody, clone 6B4 is an antibody against Aggrecan for use in Western Blotting & IHC.
Immunofluorescent Analysis: A representative lot was used by an independent laboratory in human femoral chondrocytes (Williams, R., et al. (2010). PLoS One. 5(10):e13246.).

Immunohistochemistry Analysis: A representative lot was used by an independent laboratory in human chartilage tissue (Williams, R., et al. (2010). PLoS One. 5(10):e13246.).
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated by Western Blot in A1D1 purified protein from bovine tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Aggrecan in 10 µg of purified protein from bovine tissue lysate.

Target description

~60 kDa observed. Uniprot describes three isoforms at ~250 kDa, ~246 kDa, and ~239 kDa Aggrecan is degraded by aggrecanase into a ~60 kDa protein and other higher molecular weight products (Lemons, M. L. et al. (2001). J Neurosci. 21(13):4772-4781. Forsang, A. J. et al. (1995). Biochem J. 310(Pt 1):337-343.).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A1D1 purified protein from bovine tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Cheng Lo et al.
Cell transplantation, 30, 9636897211045319-9636897211045319 (2021-09-30)
Apart from aging process, adult intervertebral disc (IVD) undergoes various degenerative processes. However, the nicotine has not been well identified as a contributing etiology. According to a few studies, nicotine ingestion through smoking, air or clothing may significantly accumulate in
Wen-Cheng Lo et al.
Life (Basel, Switzerland), 11(10) (2021-10-24)
Complications of diabetes mellitus (DM) range from acute to chronic conditions, leading to multiorgan disorders such as nephropathy, retinopathy, and neuropathy. However, little is known about the influence of DM on intervertebral disc degeneration (IVDD). Moreover, traditional surgical outcomes in
Hira Lal Goel et al.
Cell reports, 7(3), 747-761 (2014-04-29)
Although the α6β1 integrin has been implicated in the function of breast and other cancer stem cells (CSCs), little is known about its regulation and relationship to mechanisms involved in the genesis of CSCs. We report that a CD44(high)/CD24(low) population, enriched
Navneet Kumar Dubey et al.
Aging and disease, 10(3), 483-496 (2019-06-06)
Diabetes mellitus (DM) is well-known to exert complications such as retinopathy, cardiomyopathy and neuropathy. However, in recent years, an elevated osteoarthritis (OA) complaints among diabetics have been observed, portending the risk of diabetic OA. Since formation of advanced glycation end
Xiaoyi Zhao et al.
Molecular therapy. Nucleic acids, 22, 832-845 (2020-11-25)
Knee osteoarthritis (KOA) is a highly prevalent disabling joint disease in aged people. Progressive cartilage degradation is the hallmark of KOA, but its deeper mechanism remains unclear. Substantial evidence indicates the importance of the synovium for joint homeostasis. The present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service