Skip to Content
Merck
All Photos(2)

Documents

MABS518

Sigma-Aldrich

Anti-CBS Antibody, clone 9F3.2

clone 9F3.2, from mouse

Synonym(s):

Cystathionine beta-synthase, CBS, Beta-thionase, Serine sulfhydrase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9F3.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CBS(875)

General description

Cystathionine beta-synthase (EC 4.2.1.22; UniProt P35520; also known as beta-thionase, Methylcysteine synthase, Serine sulfhydrase) is encoded by the CBS (also known as HIP4) gene (Gene ID 875) in human. Cystathionine beta-synthase catalyzes the trans-sulfuration of homocysteine and serine to form the cysteine precursor cystathionine in the folate pathway. Cysteine is an essential substrate for the biosynthesis of glutathione, a major endogenous antioxidant. CBS deficiency results in altered sulfur metabolism and homocystinuria characterized by increased urinary homocystine and methionine. The clinical features of untreated homocystinuria due to CBS deficiency include myopia, ectopia lentis, mental retardation, skeletal anomalies resembling Marfan syndrome, and thromboembolic events.

Specificity

Expected to react with both the Major (isoform 1) and the Minor (isoform 2) spliced variant.

Immunogen

Recombinant protein corresponding to the N-terminal of human CBS.

Application

Anti-CBS Antibody, clone 9F3.2 is an antibody against CBS for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected CBS in human liver and human pancreas tissue.

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected CBS in 10 µg of Raji cell lysate.

Target description

~61 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service