Skip to Content
Merck

MABS1237

Anti-NKCC1/2 Antibody, clone T4

clone T4, from mouse

Synonym(s):

Solute carrier family 12 member 1, Solute carrier family 12 member 2, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1, Basolateral Na-K-Cl symporter

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
T4, monoclonal
Application:
EM, ICC, IF, IP, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

T4, monoclonal

species reactivity

gerbil, fish, canine, mouse, human, shark, bovine, avian, ferret, rat, rabbit

technique(s)

electron microscopy: suitable, immunocytochemistry: suitable, immunofluorescence: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC12A2(6558)

General description

Solute carrier family 12 member 1 (UniProt Q13621; a.k.a. Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2) and solute carrier family 12 member 2 (UniProt P55011; a.k.a. Basolateral Na-K-Cl symporter, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1) are encoded by the SLC12A1 (a.k.a. NKCC2; Gene ID 6557) and SLC12A2 (a.k.a. NKCC1; Gene ID 6558) gene, respectively, in human. NKCC1 (SLC12A2) and NKCC2 (SLC12A1) belong to the subfamily of cation-chloride cotransporters (CCCs) that mediate electroneutral transport of sodium, potassium and chloride across the plasma membrane and are inhibited by bumetanide and furosemide-type of compounds. NKCCs play an important role in cell volume regulation, where NKCC2 is involved in reabsorption of salt and osmotically-obliged water in renal epithelial cells, while NKCC1 has a major impact on intracellular chloride ion concentration ([Cl-]i) in vascular smooth muscle cells (VSMCs) and neuronal cells. In primary hypertension, NKCC1 activity is increased in VSMCs and paraventricular nucleus (PVN) neurons, leading to elevated peripheral resistance in systemic circulation and activation of sympathetic nerve system (SNS), respectively. Human NKCC1 and NKCC2 are 12-transmembrane proteins having their N- and C-terminal ends exposed at the cytoplasmic side (a.a. 1-285 and 921-1212 of NKCC1; a.a. 1-177 and 814-1099 of NKCC2).
~165 kDa observed. Target bands appear larger than the calculated molecular weights of 131.4/129.7 kDa (human NKCC1 isoform 1/2), 121.5/121.4 kDa (human NKCC2 isoform A/F), 131.0 kDa (mouse NKCC1), 120.4/120.3/120.3 kDa (mouse NKCC2 isoform A/B/F), and 130.4/120.6 kDa (rat NKCC1/NKCC2) due to glycosylation. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Recombinant human NKCC1 C-terminal fragment.

Application

Research Category
Signaling
This mouse monoclonal Anti-NKCC1/2 Antibody, clone T4, Cat. No. MABS1237 detects levels of NKCC1 & NKCC2, and has been published and validated for use in Electron Microscopy, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, and Western Blotting.

Biochem/physiol Actions

Clone T4 recognizes both NKCC1 and NKCC2 by targeting a C-terminal region epitope exposed only under denaturing condition, and unaccessible for antibody binding when NKCC assumes its native conformation (Lytle, C., et al. (1995). Am. J. Physiol. 269(6 Pt 1):C1469-C1505).

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4) 150 mM NaCl with 0.05% sodium azide

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected NKCC in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiyoshi Egawa et al.
Scientific reports, 13(1), 5685-5685 (2023-04-18)
Angelman syndrome is a neurodevelopmental disorder caused by loss of function of the maternally expressed UBE3A gene. Treatments for the main manifestations, including cognitive dysfunction or epilepsy, are still under development. Recently, the Cl- importer Na+-K+-Cl- cotransporter 1 (NKCC1) and
Trevor Sasserath et al.
Advanced therapeutics, 5(11) (2023-01-03)
There are many neurological rare diseases where animal models have proven inadequate or do not currently exist. NGLY1 Deficiency, a congenital disorder of deglycosylation, is a rare disease that predominantly affects motor control, especially control of neuromuscular action. In this

Global Trade Item Number

SKUGTIN
MABS123704054839035340

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service