Skip to Content
Merck
All Photos(1)

Key Documents

MABF2005

Sigma-Aldrich

Anti-CCR5 (CD195) Antibody, clone HEK/1/85a/7a

clone HEK/1/85a/7a, from rat

Synonym(s):

C-C chemokine receptor type 5, C-C CKR-5, CC-CKR-5, CCR-5, CCR5, CHEMR13, HIV-1 fusion coreceptor, CD195

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HEK/1/85a/7a, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CCR5(1234)

General description

C-C chemokine receptor type 5 (UniProt: P51681; also known as C-C CKR-5, CC-CKR-5, CCR-5, CCR5, CHEMR13, HIV-1 fusion coreceptor, CD195) is encoded by the CCR5 (also known as CMKBR5) gene (Gene ID: 1234) in human. CCR-5 is a multi-pass membrane protein that is expressed in memory T-cells, macrophages, and immature dendritic cells, and is upregulated by proinflammatory cytokines. It is also shown to be highly expressed in spleen, thymus, and on THP-1 and KG-1a cells. Lower expression is observed in ovary and lung. CCR-5 serves as a receptor for a number of inflammatory CC-chemokines and subsequently transduces a signal by increasing intracellular calcium levels. It also plays a role in the control of granulocytic lineage proliferation or differentiation. CCR-5 is acts as a co-receptor (CD4 being the primary receptor) of HIV-1 and HIV-2. It also couples to Gi class of G-proteins to inhibit cAMP production and stimulate calcium release. CCR-5 undergoes sulfation on two tyrosine residues in the N-terminal region, which increases efficiency of HIV-1 entry into cells. It also undergoes phosphorylation at C-terminal serine residues following binding to CC chemokines, especially APO-RANTES. Mutations in CCR5 gene have been linked to insulin-dependent diabetes mellitus. (Ref.: Blanpain, C., et al. (2002). Receptors Channels. 8(1);19-31).

Specificity

Clone HEK/1/85a/7a is a rat monoclonal antibody that specifically detects human C-C-chemokine receptor 5 (CD195).

Immunogen

CHO cells transfected with full length human CCR-5 (CD195).

Application

Anti-CCR5 (CD195), clone HEK/1/85a/7a, Cat. No. MABF2005, is a rat monoclonal antibody that detects C-C chemokine receptor type 5 (CCR5) and has been tested for use in Flow Cytometry and Immunocytochemistry.
Immunocytochemistry Analysis: A representative lot detected CCR5 (CD195) in Immunocytochemistry applications (Mueller, A., et. al. (2002). Blood. 99(3):785-91).

Flow Cytometry Analysis: A representative lot detected CCR5 (CD195) in Flow Cytometry applications (Mueller, A., et. al. (2002). Blood. 99(3):785-91).

Quality

Evaluated by Flow Cytometry in wrote RBL-2H3 cells stably transfection with CCR5.

Flow Cytometry Analysis (FC): 1 µg of this antibody detected C-C chemokine receptor type 5 (CD195) in one million CCR-5 transfected RBL-2H3 cells.

Target description

40.52 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service