Skip to Content
Merck
All Photos(1)

Key Documents

MABE867

Sigma-Aldrich

Anti-Mad1 Antibody, clone BB3-8

clone BB3-8, from mouse

Synonym(s):

Mitotic spindle assembly checkpoint protein MAD1, Mitotic arrest deficient 1-like protein 1, MAD1-like protein 1, Mitotic checkpoint MAD1 protein homolog, HsMAD1, hMAD1, Tax-binding protein 181

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BB3-8, monoclonal

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAD1L1(8379)

General description

Mitotic arrest deficient-like 1 (yeast) is also known as MAD1 and MAD1L1. It is part of the MAD1 family which is crucial to development. MAD1 acts as a checkpoint during mitotic spindle assembly. During anaphase and telophase MAD1 is located at the spindle mid-zone of the cell, where it prevents anaphase until the chromosome is aligned at the metaphase plate. During metaphase MAD1 is located at the centrosome. MAD1 participates in cell cycle control and tumor suppression. MAD1 functions as a homodimer and interacts with MAD2L1. It is thought that MAD1 recruits to MAD2, which then promotes binding of MAD2 to CDC20.

Immunogen

His-tagged recombinant protein corresponding to human Mad1.

Application

Anti-Mad1 Antibody, clone BB3-8 is a highly specific mouse monoclonal antibody, that targets MAD1 & has been tested in western blotting & Immunofluorescence.
Immunofluorescence Analysis: A representative lot from an independent laboratory detected Mad1 in HeLa cells (Santaguida, S., et al. (2011). EMBO J. 30(8):1508-1519.).

Immunoflourescence Analysis: A representative lot from an independent laboratory detected Mad1 in HeLa cells (Screpanti, E., et al. (2011). Curr Biol. 21(5):391-398.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Mad1 in HeLa cell lysate (Screpanti, E., et al. (2011). Curr Biol. 21(5):391-398.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Mad1 in 10 µg of HeLa cell lysate.

Target description

~83 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Corno et al.
The EMBO journal, 42(20), e112630-e112630 (2023-09-15)
Two major mechanisms safeguard genome stability during mitosis: the mitotic checkpoint delays mitosis until all chromosomes have attached to microtubules, and the kinetochore-microtubule error-correction pathway keeps this attachment process free from errors. We demonstrate here that the optimal strength and
Shawn Yost et al.
Nature genetics, 49(7), 1148-1151 (2017-05-30)
Through exome sequencing, we identified six individuals with biallelic loss-of-function mutations in TRIP13. All six developed Wilms tumor. Constitutional mosaic aneuploidies, microcephaly, developmental delay and seizures, which are features of mosaic variegated aneuploidy (MVA) syndrome, were more variably present. Through
Jingchao Wu et al.
The Journal of cell biology, 223(1) (2023-11-07)
Correct chromosome segregation during cell division depends on proper connections between spindle microtubules and kinetochores. During prometaphase, kinetochores are temporarily covered with a dense protein meshwork known as the fibrous corona. Formed by oligomerization of ROD/ZW10/ZWILCH-SPINDLY (RZZ-S) complexes, the fibrous
Adrian T Saurin et al.
Methods in molecular biology (Clifton, N.J.), 1413, 333-347 (2016-05-20)
Mitotic kinetochores are signaling network hubs that regulate chromosome movements, attachment error-correction, and the spindle assembly checkpoint. Key switches in these networks are kinases and phosphatases that enable rapid responses to changing conditions. Describing the mechanisms and dynamics of their
A kinesin-based approach for inducing chromosome-specific mis-segregation in human cells.
Truong, et al.
The Embo Journal, 42, e111559-e111559 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service