Skip to Content
Merck

MABC602

Anti-Ferritin Heavy Chain Antibody, clone 1-2.3.1.2

clone 1-2.3.1.2, from mouse

Synonym(s):

Ferritin heavy chain, Ferritin H subunit, Cell proliferation-inducing gene 15 protein, Ferritin heavy chain, N-terminally processed

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1-2.3.1.2, monoclonal
Application:
IHC, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1-2.3.1.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FTH1(2495)

General description

Ferritin is a widely expressed, very large, multi-subunit, intracellular protein that stores iron and releases it in a controlled fashion. The protein is produced by almost all living organisms in some form. In humans, it acts as a buffer against iron deficiency and iron overload. Ferritin protein is a multi-subunit (24 oligomers) protein complex consisting of both large (heavy) and small (light) chains. It is primarily and intracellular protein.The subunits of Ferritin in vertebrates have an apparent molecular weight of 19 kDa or 21 kDa respectively; their sequences are about 50% homologous. When Ferritin is unbound by iron the protein is called Apoferritin. Mutations associated with the Ferritin Heavy Chain protein contribute to a syndrome called Hemochromatosis 5 which is a disorder of iron metabolism where too much iron is retained within the tissues resulting in a variety of syndromes which unfortunately manifest themselves late in life.
~21 kDa observed. Uncharacterized band may be observed at ~200 kDa

Immunogen

Recombinant protein corresponding to human Ferritin Heavy Chain.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Ferritin Heavy Chain in human bone marrow and human tonsil tissue.
Immunohistochemistry Analysis: A representative lot detected Ferritin Heavy Chain in erythroid blasts of acute myeloid leukaemia M6 tissue, macrophages in the lymph node tissue, and tumor & lung-associated macrophages (Wang, W., et al. (2013). Histopahtology. 62:931–940).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Ferritin Heavy Chain Antibody, clone 1-2.3.1.2 is validated for use in Western Blotting and Immunohistochemistry for the detection of Ferritin Heavy Chain.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in U251 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Ferritin Heavy Chain in 10 µg of U251 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library