Skip to Content
Merck

MABC147

Anti-TRAIL/CD253 Antibody, clone 6D12.2

clone 6D12.2, from mouse

Synonym(s):

Tumor necrosis factor ligand superfamily member 10, Apo-2 ligand, Apo-2L, TNF-related apoptosis-inducing ligand, Protein TRAIL, CD253

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
6D12.2, monoclonal
Application:
FACS, IHC
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6D12.2, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable, immunohistochemistry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TNFSF10(8743)

General description

33 kDa calculated
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 and Fas. A novel member in the TNF family was recently identified and designated TRAIL (for TNF-related apoptosis-inducing ligand) and Apo-2L (for Apo-2 ligand). TRAIL is a type II membrane protein and is expressed in a variety of human tissues. Two novel death domain containing receptors DR4 and DR5 have been identified as the receptor for TRAIL. Like TNF and Fas ligand, TRAIL induces apoptosis and NF-kB activation in many tissues and cells.

Immunogen

GST-tagged recombinant protein corresponding to human TRAIL/CD253.

Application

Anti-TRAIL/CD253 Antibody, clone 6D12.2 is an antibody against TRAIL/CD253 for use in flow cytometry & IHC.
Flow Cytometry Analysis: 4 µg of this antibody detected TRAIL/CD253 in 1X10E6 human PBMC, which were treated with a proinflammatory stimulus.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected TRAIL/CD253 in human prostate cancer sections. (Note: This antibody detects TRAIL/CD253 in cancer tissues and not in normal tissues.).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Flow Cytometry in HeLa cells, which were treated with a proinflammatory stimulus.

Flow Cytometry Analysis: 4 µg of this antibody detected TRAIL/CD253 in 1X10E6 HeLa cells, which were treated with a proinflammatory stimulus.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MABC14704053252955136