Skip to Content
Merck
All Photos(2)

Key Documents

MAB5360

Sigma-Aldrich

Anti-Spinocerebellar Ataxia Type 3 Antibody, clone 1H9

ascites fluid, clone 1H9, Chemicon®

Synonym(s):

Ataxin-3, josephin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1H9, monoclonal

species reactivity

rat, human, monkey, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ATXN3(4287)

General description

Spinocerebellar ataxia (SCA) is a genetic disease with multiple types, each of which could be considered a disease in its own right.
Spinocerebellar ataxia (SCA) is one of a group of genetic disorders characterized by slowly progressive incoordination of gait and often associated with poor coordination of hands, speech, and eye movements. Frequently, atrophy of the cerebellum occurs. The first ataxia gene was identified in 1993 for a dominantly inherited type called “Spinocerebellar ataxia type 1" (SCA1). Subsequently, as additional dominant genes were found they were called SCA2, SCA3, etc. Usually, the "type" number of "SCA" refers to the order in which the gene was found. At this time, there are at least 29 different gene mutations which have been found.

Specificity

Ataxin-3. The epitope was mapped precisely at E214-L233. MAB5360 can be used to study wild type ataxin-3 and the mutant form with polyglutamine expansion found in patients affected with spinocerebellar ataxin type 3/Machado-Joseph disease (SCA3/MJD). In analysis of human tissues by Western blot, MAB5360 releaved several isoforms of ataxin-3 (presumably generated by alternative splicing, Trottier et al. 1998). The antibody detected polyglutamine aggregate (or nuclear inclusions) by IHC on SCA-3/MJD brain sections (Paulson et al. 1997).

Immunogen

Human ataxin-3 fragment from aa F112-L249 as a fusion protein

Application

Detect Spinocerebellar Ataxia Type 3 using this Anti-Spinocerebellar Ataxia Type 3 Antibody, clone 1H9 validated for use in ELISA, IC, IH, IP & WB.
Immunohistochemistry:
A 1:500-1:5000 dilution of a previous lot was used in IH.

Immunoprecipitation:
A 1:500-1:5000 dilution of a previous lot was used in IP.

ELISA:
A 1:500-1:5000 dilution of a previous lot was used in ELISA.

Immunocytochemistry:
A 1:500-1:5000 dilution of a previous lot was used in IC.

Optimal working dilutions must be determined by the end user.

Quality

Evaluated by Western Blot on NIH/3T3 lysates.

Western Blot Analysis:
1:500 dilution of this antibody detected SPINOCEREBELLAR ATAXIA 3 on 10 µg of NIH/3T3 lysates.

Target description

44 kDa

Analysis Note

Control
Human SCA-3/MJD brain sections, NIH/3T3 lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huu Phuc Nguyen et al.
PloS one, 8(4), e62043-e62043 (2013-04-30)
Spinocerebellar Ataxia Type 3 (SCA3), also known as Machado-Joseph disease, is an autosomal dominantly inherited neurodegenerative disease caused by an expanded polyglutamine stretch in the ataxin-3 protein. A pathological hallmark of the disease is cerebellar and brainstem atrophy, which correlates
Sandro Alves et al.
Human molecular genetics, 17(14), 2071-2083 (2008-04-04)
Machado-Joseph disease (MJD) is a fatal, dominant neurodegenerative disorder. MJD results from polyglutamine repeat expansion in the MJD-1 gene, conferring a toxic gain of function to the ataxin-3 protein. In this study, we aimed at overexpressing ataxin-3 in the rat
Jiaxin Hu et al.
Nature biotechnology, 27(5), 478-484 (2009-05-05)
Expanded trinucleotide repeats cause many neurological diseases. These include Machado-Joseph disease (MJD) and Huntington's disease (HD), which are caused by expanded CAG repeats within an allele of the ataxin-3 (ATXN3) and huntingtin (HTT) genes, respectively. Silencing expression of these genes
Isabel Onofre et al.
Scientific reports, 6, 28220-28220 (2016-06-23)
Machado Joseph Disease (MJD) is the most frequent autosomal dominantly inherited cerebellar ataxia caused by the over-repetition of a CAG trinucleotide in the ATXN3 gene. This expansion translates into a polyglutamine tract within the ataxin-3 protein that confers a toxic
Pawel M Switonski et al.
Neuromolecular medicine, 13(1), 54-65 (2010-10-15)
Spinocerebellar ataxia 3 (SCA3) is a genetic disorder resulting from the expansion of the CAG repeats in the ATXN3 gene. The pathogenesis of SCA3 is based on the toxic function of the mutant ataxin-3 protein, but the exact mechanism of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service