Skip to Content
Merck
All Photos(4)

Key Documents

MAB353

Sigma-Aldrich

Anti-Nestin Antibody, clone rat-401

clone rat-401, Chemicon®, from mouse

Synonym(s):

rat401

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

rat-401, monoclonal

species reactivity

mouse, rat

should not react with

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

input

sample type neural stem cell(s)
sample type epithelial cells

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NES(10763)
mouse ... Nes(18008)
rat ... Nes(25491)

General description

Nestin, a large intermediate filament protein (class Type VI) expressed during development and in myotendinous and neuromuscular junctions. Nestin expression is restricted, typically disappearing by E18. Nestin is thought to be a reasonable neuronal marker; however, recent studies have found nestin expression in other cell types such as endothelial cells (Mokry and Nemecek, 1998). Nestin identifies the most primitive neuroepithelium but also identifies many other embryonic tissues, so it is not specific for CNS. Nestin expression is seen in almost all GBMs (Glioblastoma mulitformes) and many melanomas (both primary and metastatic) but not in any metastatic carcinomas.

Specificity

Rodent Nestin protein confirmed with rat and mouse. Rat-401 is not reactive with human nestin protein.

Immunogen

Nestin purified from embryonic rat spinal cord.

Application

Detect Nestin using this Anti-Nestin Antibody, clone rat-401 validated for use in IC, IH, IH(P) & WB.
Research Category
Stem Cell Research
Research Sub Category
Neural Stem Cells
Western Blot:
200-220 kDa in reducing gels of newborn rat or mouse cell extracts. Note that nestin was originally reported (erroneously) to migrate at approximately 440 kDa by SDS-PAGE (Kachinsky, 1995).
For Westerns, prepare samples by boiling in 4 volumes of 125 mM Tris, pH 6.8, 10% 2-mercaptoethanol, 10% glycerol and 4.6% SDS. 5% PAGE gels are suggested. Block membranes with milk or BSA. Do not dilute primary antibody in buffer containing milk. Freezing and thawing of samples or samples not stored at -85°C will show breakdown of nestin sometimes into multiple fragments.

Immunohistochemistry:
1:20-1:200 dilution of a previous lot was used. Perfuse tissue with 4% paraformaldehyde at pH 7.4 for light microscopy or with either 4% paraformaldehyde at pH 10.0 or 4% paraformaldehyde with 0.1% glutaraldehyde at pH 7.4 for EM. Suitable for frozen or paraffin-embedded sections.

Immunocytochemistry:
1:20-1:200 dilution of a previous lot was used. Fix cells in 4% paraformaldehyde buffered with 50 mM sodium borate at pH 9.5.

Optimal working dilutions and protocols must be determined by end user.

Quality

Immunohistochemistry(paraffin):
Nestin (cat. # MAB353, Clone 401) staining of Mouse Pancreas, tissue was pretreated with citrate buffer, pH 6.0. A previous lot of this antibody was diluted to 1.25 µg/mL, using IHC-Select detection with HRP-DAB. Immunoreactivity is detected in the islets of Langerhans.
Optimal Staining of Nestin Monoclonal Antibody: Mouse Pancreas

Target description

200-220 kDa

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG1 in buffer containing 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Mouse embryonic brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Wang et al.
Stem cells (Dayton, Ohio), 30(2), 169-179 (2011-12-02)
Embryonic stem cell (ESC)-based therapy is a promising treatment for neurodegenerative diseases. But there is always a risk of tumor formation that is due to contamination of undifferentiated ESCs. To reduce the risk and improve ESC-based therapy, we have established
Matthew C Holman et al.
Investigative ophthalmology & visual science, 51(4), 2197-2207 (2009-10-31)
Purpose. Because of differences in energy metabolism between the brain and retina, the hypothesis for the study was that, in a model of ocular and cerebral hypoperfusion, the retina would be protected by short-term hyperglycemia, whereas brain injury would be
Neuronatin promotes neural lineage in ESCs via Ca(2+) signaling.
Lin, HH; Bell, E; Uwanogho, D; Perfect, LW; Noristani, H; Bates, TJ; Snetkov, V; Price, J; Sun, YM
Stem Cells null
Bradley W Doble et al.
Developmental cell, 12(6), 957-971 (2007-06-05)
In mammalian cells, glycogen synthase kinase-3 (GSK-3) exists as two homologs, GSK-3alpha and GSK-3beta, encoded by independent genes, which share similar kinase domains but differ substantially in their termini. Here, we describe the generation of an allelic series of mouse
Isoflurane inhibits growth but does not cause cell death in hippocampal neural precursor cells grown in culture.
Sall, JW; Stratmann, G; Leong, J; McKleroy, W; Mason, D; Shenoy, S; Pleasure, SJ; Bickler, PE
Anesthesiology null

Articles

Human iPSC neural differentiation media and protocols used to generate neural stem cells, neurons and glial cell types.

Human iPSC neural differentiation media and protocols used to generate neural stem cells, neurons and glial cell types.

Human iPSC neural differentiation media and protocols used to generate neural stem cells, neurons and glial cell types.

Human iPSC neural differentiation media and protocols used to generate neural stem cells, neurons and glial cell types.

Protocols

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service