Skip to Content
Merck
All Photos(1)

Key Documents

MAB3482

Sigma-Aldrich

Anti-Cystic Fibrosis Transmembrane Conductance Regulator Antibody, clone MM13-4

clone MM13-4, Chemicon®, from mouse

Synonym(s):

CFTR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MM13-4, monoclonal

species reactivity

human

should not react with

mouse, shark

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CFTR(1080)

Specificity

Human cystic fibrosis transmembrane conductance regulator (CFTR). Recognizes an epitope at the N-terminal between residues 24 and 35.

Immunogen

Synthetic peptide (RKGYRQRLELSD) corresponding to residues 25-36 of human cystic fibrosis transmembrane conductance regulator (CFTR).

Application

Detect Cystic Fibrosis Transmembrane Conductance Regulator using this Anti-Cystic Fibrosis Transmembrane Conductance Regulator Antibody, clone MM13-4 validated for use in IC, IP & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 10 μg/mL, recognizes CFTR (150-170kDa) and two additional proteins at 52kDa and 38kDa

Immunoprecipitation

Immunofluorescence

Note: Does not work on paraffin embedded tissue.

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02 M Phosphate buffer, 0.25 M NaCl, pH 7.6 with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunying Li et al.
The Journal of biological chemistry, 279(23), 24673-24684 (2004-04-03)
Based on electrophysiological measurements, it has been argued that the active form of cystic fibrosis trans-membrane conductance regulator (CFTR) Cl(-) channel is a multimer. It has also been demonstrated that this multimerization is likely due to PDZ domain-interacting partners. Here
Loïc Drévillon et al.
PloS one, 6(3), e18334-e18334 (2011-04-13)
The CFTR (cystic fibrosis transmembrane conductance regulator) protein is a large polytopic protein whose biogenesis is inefficient. To better understand the regulation of CFTR processing and trafficking, we conducted a genetic screen that identified COMMD1 as a new CFTR partner.
Jie Ting Zhang et al.
Scientific reports, 7(1), 5233-5233 (2017-07-14)
Cystic fibrosis transmembrane conductance regulator (CFTR), known as a cAMP-activated Cl- channel, is widely expressed at the apical membrane of epithelial cells in a wide variety of tissues. Of note, despite the abundant expression of CFTR in mammalian kidney, the
Hong Yu Ren et al.
Molecular biology of the cell, 24(19), 3016-3024 (2013-08-09)
Cystic fibrosis (CF) is a fatal genetic disorder associated with defective hydration of lung airways due to the loss of chloride transport through the CF transmembrane conductance regulator protein (CFTR). CFTR contains two membrane-spanning domains (MSDs), two nucleotide-binding domains (NBDs)
Iwona Pranke et al.
ERJ open research, 4(1) (2018-03-03)
Premature termination codons (PTCs) are generally associated with severe forms of genetic diseases. Readthrough of in-frame PTCs using small molecules is a promising therapeutic approach. Nonetheless, the outcome of preclinical studies has been low and variable. Treatment efficacy depends on:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service