Skip to Content
Merck
All Photos(1)

Key Documents

MAB3319

Sigma-Aldrich

Anti-MMP-14 Antibody, catalytic domain, clone 114-6G6

clone 114-6G6, Chemicon®, from mouse

Synonym(s):

MT1-MMP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

114-6G6, monoclonal

species reactivity

human

should not react with

pig, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MMP14(4323)

Specificity

The antibody specifically reacts with human MT1-MMP. This is a purified mouse monoclonal antibody to an oligopeptide of REVPYAYIREGHEK (residue 160-173) on human membrane-type 1 matrix metalloproteinase (human MT1-MMP) and does not react with mouse or rat. However, this sequence is identical in mouse and rat MT1-MMP. For unknown reasons this antibody does not react on westerns to CC1043.

Immunogen

Epitope: catalytic domain

Application

Immunoblotting: 8-20 μg/mL. 66+60kDa (Ueno et al., 1997)

Immunohistochemistry on paraffin-embedded tissue sections, use at 20 μg/mL concentration (see Ueno et al., 1997; Nakamura et al., 1999). Requires periodate-lysine-paraformaldehyde fixative (Namura et al., 1999) for 18-24 hrs. at 4°C.

EIA Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
This Anti-MMP-14 Antibody, catalytic domain, clone 114-6G6 is validated for use in ELISA, WB, IH(P) for the detection of MMP-14.

Physical form

Format: Purified
Liquid in 0.1 M sodium phosphate buffer, pH 7.0 containing 2% protease free bovine serum albumin.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Manufactured by Daiichi Fine Chemical Co., Ltd

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongping Li et al.
American journal of translational research, 7(1), 120-127 (2015-03-11)
Membrane type 1 matrix metalloproteinase (MT1-MMP) has been demonstrated to play an important role in tumor progression. The aim of the present study was to analyze the expression of MT1-MMP in breast cancer and its correlation with clinicopathologic characteristics, including
Bo-Ra Son et al.
Stem cells (Dayton, Ohio), 24(5), 1254-1264 (2006-01-18)
Human mesenchymal stem cells (MSCs) are increasingly being considered in cell-based therapeutic strategies for regeneration of various organs/tissues. However, the signals required for their homing and recruitment to injured sites are not yet fully understood. Because stromal-derived factor (SDF)-1 and
Dae-Il Chang et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 23(12), 1408-1419 (2003-12-10)
During focal cerebral ischemia, matrix metalloproteinase-2 (MMP-2) can contribute to the loss of microvessel integrity within ischemic regions by degrading the basal lamina. MMP-2 is secreted in latent form (pro-MMP-2), but the activation of pro-MMP-2 in the ischemic territory has
Anna Janowska-Wieczorek et al.
International journal of cancer, 113(5), 752-760 (2004-10-23)
The role of platelets in tumor progression and metastasis has been recognized but the mechanism of their action remains unclear. Five human lung cancer cell lines (A549, CRL 2066, CRL 2062, HTB 183, HTB 177) and a murine Lewis lung
David H Lovett et al.
Biochemical and biophysical research communications, 398(3), 482-488 (2010-07-06)
YB-1 is a member of the cold shock domain family, with complex roles in DNA structure, gene transcription and translation. YB-1 promotes chromosomal instability, and mammary gland transgenic expression induces tumors with 100% penetrance. YB-1 is linked to poor prognosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service