Skip to Content
Merck
All Photos(1)

Key Documents

MAB2146

Sigma-Aldrich

Anti-ICAM-1 Antibody, clone P2A4

clone P2A4, Chemicon®, from mouse

Synonym(s):

CD54

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

P2A4, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ICAM1(3383)

Related Categories

Specificity

Human ICAM-1

Immunogen

IL-1beta activated HUVEC (human)

Application

Anti-ICAM-1 Antibody, clone P2A4 is an antibody against ICAM-1 for use in ELISA, FC, IP, IC, IH.
Immunohistochemistry: for use on acetone or paraformaldehyde fixed tissue

Immunocytochemistry: for use on B cells, PBL, T cells

Immunoprecipitation

FACS

ELISA

Inhibits ICAM-1 dependent adhesive interactions

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Physical form

Format: Purified
Nonsterile liquid contains 0.2M phosphate buffer, 0.25M sodium chloride, and 0.1% sodium azide. Filtered through a 0.45 micron membrane filter.

Storage and Stability

Maintain at +2-8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alba Carmona et al.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation, 19(10), 1439-1445 (2013-07-13)
Endothelial dysfunction seems to be a key factor in the development of several complications observed early after hematopoietic stem cell transplantation (HSCT). The conditioning regimen and many other factors associated with the procedure are responsible for this endothelial damage. The
João M Furtado et al.
Immunology and cell biology, 90(9), 912-915 (2012-04-25)
Retinal infection is the most common clinical manifestation of toxoplasmosis. The route by which circulating Toxoplasma gondii tachyzoites cross the vascular endothelium to enter the human retina is unknown. Convincing studies using murine encephalitis models have strongly implicated leukocyte taxis
E Pluskota et al.
The Journal of biological chemistry, 275(39), 30029-30036 (2000-06-24)
Intercellular adhesion molecule-1 (ICAM-1) binds to the plasma protein fibrinogen (Fg) to mediate leukocyte/endothelial cell interactions. In our studies, the ligation of Fg to ICAM-1 on tumor necrosis factor-alpha-stimulated endothelial cells resulted in the tyrosine phosphorylation of Src homology domain
Akio Kawakami et al.
Circulation, 114(7), 681-687 (2006-08-09)
Activation of vascular endothelial cells (ECs) plays an important role in atherogenesis and plaque instability. Lipoproteins containing apolipoprotein CIII (apoCIII) predict coronary heart disease (CHD). We recently reported that apoCIII has a proinflammatory effect on human monocytes. In this study
Qiangjun Cai et al.
Arteriosclerosis, thrombosis, and vascular biology, 24(12), 2263-2270 (2004-10-02)
Vascular smooth muscle cells (VSMCs) may regulate monocyte functions within atherosclerotic lesions. We investigated the impact of VSMC/monocyte interactions on monocyte apoptosis and scavenger receptor CD36 expression, key events related to monocyte survival and differentiation. Serum deprivation significantly increased THP-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service