Skip to Content
Merck
All Photos(1)

Key Documents

MAB2081

Sigma-Aldrich

Anti-β-Catenin Antibody, clone 5H10

clone 5H10, Chemicon®, from mouse

Synonym(s):

Anti-CTNNB, Anti-EVR7, Anti-MRD19, Anti-NEDSDV, Anti-armadillo

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5H10, monoclonal

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CTNNB1(1499)
rat ... Ctnnb1(84353)

General description

The transmembrane, calcium dependent molecule E-cadherin is considered to be one of the key molecules in the formation of the intercellular junctional complex and establishment of polarity in epithelial cells. Catenins, the alpha-subunit (102 kDa), beta-subunit (88 kDa) and gamma-subunit (82 kDa), [reviewed in Takeichi, 1991] are a group of proteins that interact with the intercellular domain of E-cadherin, resulting in complexes of E-cadherin/beta-catenin/alpha-catenin or E-cadherin/gamma-catenin/alpha-Catenin (Hinck et al., 1994). Beta-catenin, the vertebrate homologue of Drosophila segment polarity gene armadillo, and key element of the Wnt signal transduction pathways, is important in embryogenesis, cell transformation, and intercellular adhesion including the adhesion, motility and metastasis of cancer cells. Thus, catenin protein expression, regulation and localization are potentially important markers to predict motility and invasiveness of epithelial neoplasms. The regions of both alpha- and beta-catenin, located on 5q21-22 and 3p21, have been shown to be involved in the development of certain tumors (see Jiang, 1996; Schlosshauer et al., 2002) and reduced expressions of both alpha- and beta-catenin have been described in various tumors including breast carcinoma (Hashizume et al, 1996). Besides adhesion functions, beta-catenin binds to the tumor suppressor gene APC. APC mutation disturbs the equilibrium and cytoplasmic levels of free beta-catenin level in the cell and may have a role in tumorigenesis.

Specificity

Recognizes an epitope in the carboxy terminus, amino acids 758-771, of human beta-catenin.

Immunogen

Produced by immunization with recombinant human beta-catenin.

Application

Anti-β-Catenin Antibody, clone 5H10 is a Mouse Monoclonal Antibody for detection of β-Catenin also known as beta catenin & has been tested in ICC, IHC, IHC(P), IP & WB.
Immunohistochemistry: Low cross-linking fixatives are recommended. Histochoice fixative is effective. Formalin and PFA are effective, however may show weaker staining.

Immunohistochemistry on formalin fixed paraffin embedded tissues requires light fixations and citric acid/microwave antigen recovery for staining; In brief, sections of 2μm thickness were treated after deparaffinization with 0.3% hydrogen peroxide in methanol for 30 minutes to inhibit endogenous peroxidase and washed with TBS solution (Tris-buffered saline pH 7.6). To enhance antigen retrieval, sections were microwave-pretreated in 0.01 M citrate buffer solution at 750 W for a 3 minute-cycle period repeated three times; amplified detection methods are recommended.

Immunocytochemistry: Effective for A431 and HT1080 cell lines with acetone or methanol, or light PFA fixation.

Immunoblotting

Immunoprecipitation

Optimal working dilutions must be determined by end-user.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Target description

94 kDa

Linkage

Replaces: 04-1002

Physical form

Format: Purified
Protein A Purified IgG in liquid at 1 mg/mL in 0.02M PB with 0.25M NaCl, pH 7.6, containing 0.1% sodium azide.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
A431 and HT1080 cell line extracts

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cong Tong et al.
Cell biology international, 44(10), 2075-2085 (2020-07-12)
DNA-binding protein A (dbpA) is reported to be upregulated in many cancers and associated with tumor progress. The present study aimed to investigate the role of dbpA in 5-fluorouracil (5-FU)-resistant and oxaliplatin (L-OHP)-resistant colorectal cancer (CRC) cells. We found that
Qinggui Liu et al.
Aging and disease, 12(5), 1238-1251 (2021-08-04)
Previous study showed that senescent hepatocytes from aged liver could be rejuvenated after repopulated in the young recipient liver. The proliferative capacity of hepatocytes was restored with the senescence reversal. However, it is unknown whether metabolic and homeostatic function of
Noelle Leary et al.
Journal of extracellular vesicles, 11(2), e12197-e12197 (2022-02-22)
Tumour-draining lymph nodes (LNs) undergo massive remodelling including expansion of the lymphatic sinuses, a process that has been linked to lymphatic metastasis by creation of a pre-metastatic niche. However, the signals leading to these changes have not been completely understood.
Sarah Klein et al.
Journal of cell science, 129(13), 2573-2585 (2016-05-21)
Lymphangiogenesis plays a crucial role during development, in cancer metastasis and in inflammation. Activation of VEGFR-3 (also known as FLT4) by VEGF-C is one of the main drivers of lymphangiogenesis, but the transcriptional events downstream of VEGFR-3 activation are largely
Cristina Belgiovine et al.
International journal of oncology, 39(5), 1199-1204 (2011-07-14)
Studying neoplastic transformation of telomerase immortalized human fibroblasts (cen3tel), we found that the transition from normal to tumorigenic cells was associated with the loss of growth contact inhibition, the acquisition of an epithelial-like morphology and a change in actin organization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service