Skip to Content
Merck
All Photos(4)

Key Documents

MAB1950

Sigma-Aldrich

Anti-Integrin α2 Antibody, clone P1E6

clone P1E6, Chemicon®, from mouse

Synonym(s):

CD49b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
P1E6, monoclonal
application:
ICC
IHC
species reactivity:
human
technique(s):
immunocytochemistry: suitable
immunohistochemistry: suitable
citations:
63

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P1E6, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1

suitability

not suitable for immunohistochemistry (Paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA2(3673)

Specificity

Reacts with Human a2 integrin.

Application

Anti-Integrin α2 Antibody, clone P1E6 is an antibody against Integrin α2 for use in IC, IH.
Research Category
Cell Structure
Research Sub Category
Integrins
Suitable for use in attachment inhibition assays using fibroblasts, epithelial cells, endothelial cells, and non-activated platelets on collagen types I, III, IV, VI and laminin.

Suitable for immunofluorescence using fresh frozen or acetone fixed tissues. Not recommended for traditional formalin fixed paraffin embedded tissues.

Final working dilutions must be determined by end user.

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 0.01 M phosphate buffered saline, pH 7.1, with 0.1% sodium azide as a preservative.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Skin (Basement membrane)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P D Arora et al.
The American journal of pathology, 154(3), 871-882 (1999-03-18)
Wound contraction is mediated by myofibroblasts, specialized fibroblasts that appear in large numbers as the wound matures and when resistance to contractile forces increases. We considered that the regulation of myofibroblast differentiation by wound-healing cytokines may be dependent on the
P Sriramarao et al.
Endothelium : journal of endothelial cell research, 4(2), 85-97 (1996-01-01)
A direct correlation was observed between the invasive and metastatic potential of A375 melanoma cells and their expression of tenascin and ability to support endothelial cell adhesion and reorganization. Since the ability to metastasize and establish a neovasculature requires interaction
Hirokazu Date et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 28(2), 225-231 (2009-09-03)
Cellular activities responding to growth factors are important in ligament healing. The anterior cruciate ligament (ACL) has poor healing potential compared to the medial collateral ligament (MCL). To assess the differences, we investigated the proliferation, migration, adhesion, and matrix synthesis
B K Pilcher et al.
The Journal of cell biology, 137(6), 1445-1457 (1997-06-16)
We have shown in a variety of human wounds that collagenase-1 (MMP-1), a matrix metalloproteinase that cleaves fibrillar type I collagen, is invariably expressed by basal keratinocytes migrating across the dermal matrix. Furthermore, we have demonstrated that MMP-1 expression is
Anandi Rajan et al.
Scientific reports, 8(1), 10019-10019 (2018-07-04)
The enteric species F human adenovirus types 40 and 41 (HAdV-40 and -41) are the third most common cause of infantile gastroenteritis in the world. Knowledge about HAdV-40 and -41 cellular infection is assumed to be fundamentally different from that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service