Skip to Content
Merck

MAB030

Anti-DNA Antibody, double stranded, clone BV16-13

culture supernatant, clone BV16-13, Chemicon®

Synonym(s):

Double-stranded DNA antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
BV16-13, monoclonal
Technique(s):
ELISA: suitable, immunocytochemistry: suitable, radioimmunoassay: suitable
Application:
ELISA, ICC, RIA
Citations:
30
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

culture supernatant

antibody product type

primary antibodies

clone

BV16-13, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, immunocytochemistry: suitable, radioimmunoassay: suitable

isotype

IgG2aκ

shipped in

wet ice

target post-translational modification

unmodified

Immunogen

Epitope: double stranded, 28% reactive with single stranded

Application

Detect DNA with Anti-DNA Antibody, double stranded, clone BV16-13 (Mouse Monoclonal Antibody), that has been shown to work in ELISA, RIA, ICC.
EIA/RIA

Immunocytochemistry

Optimal working dilutions must be determined by end user.
Research Category
Epigenetics & Nuclear Function

Apoptosis & Cancer
Research Sub Category
Cell Cycle, DNA Replication & Repair

Apoptosis - Additional

Biochem/physiol Actions

Double stranded DNA (dsDNyA) (B-form) from human. Shows 95% reactivity to dsDNA and 28% reactivity to ssDNA.

BINDING ASSAY:

Supernatant was incubated with 32P-end labeled heat-denatured (ss)DNA or duplex (ds)DNA restriction fragments (20 pmoles nucleotides; 16,000 cpm) followed by precipitation of anti-DNA/DNA complexes with excess rabbit anti-mouse and quantitation of bound [32P] DNA by Chernkov radiation.



ANTIGEN BINDING

PROFILE: Relative Binding To:



Natural DNA Synthetic homopolymers

dsDNA(a) ssDNA(b) poly(dA) poly(dC) poly(dG) poly(dT) poly(dI) poly(dU)

1.00 0.45 0.01 0.01 0.01 0.01 0.01 0.01



Nucleic acids were complexed with solid phase mBSA and incubated with anti-DNA antibody. Radioiodinated protein A binding at the highest antibody quantity tested (either 10 mg of purified antibody or a 1:2 culture supernatant dilution) was measured and normalized relative to that determined for the preferred antigen (given a value of 1.0).



a) Native calf thymus DNA b) Heat-denatured calf thymus DNA

Physical form

Unpurified tissue culture supernatant from a perfusion system, filtered through a 0.2μ micron membrane prior to vialing. Product contains 20%FBS and Ciprofloxacin at final concentration of 10μg/mL.

Preparation Note

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library