LSKMAGH
PureProteome Nickel Magnetic Bead System
PureProteome Nickel Magnetic Beads provide researchers with a powerful bead system to help purify polyhistidine-tagged recombinant proteins
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41105507
eCl@ss:
32160405
NACRES:
NA.56
Recommended Products
conjugate
magnetic beads conjugate
form
liquid
packaging
pkg of 10 mL
manufacturer/tradename
PureProteome
technique(s)
protein purification: suitable (histidine tagged recombinant protein)
particle size
10 μm
capacity
1-5.5 mg/mL, bead suspension binding capacity (protein)
shipped in
ambient
General description
PureProteome™ magnetic bead purification systems ensure extremely fast and easy protein purification including those polyhistidine-tagged recombinant proteins. The systems feature low nonspecific binding and minimal sample loss. PureProteome Nickel Magnetic Beads provide comparable yield while offering high purities than other magnetic purification systems.
Application
Research Category
All
All
Legal Information
PureProteome is a trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service