Skip to Content
Merck
All Photos(1)

Key Documents

GG-PACK

Millipore

Bulk Reagent - ABO & RHD (Rh) Specificities

CORE Anti-D (clone RUM-1) Bulk Reagent plus Diluent Kit

Synonym(s):

Anti-D (clone RUM-1) Blood Grouping Reagent plus Diluent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102
NACRES:
NA.41

biological source

human

antibody form

unpurified

clone

RUM-1, monoclonal

form

liquid

packaging

kit of

concentration

25-40 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

6.6-7.0

isotype

IgM

shipped in

ambient

storage temp.

2-8°C

General description

Appropriate for patient and neonatal testing - does not detect DVI red cellsContains 0.1% (w/v) Sodium AzideFiltered through a 0.22μm terminal filterShelf Life: 36 months from date of filtration

Application

For use by Tube;Slide;Microplate;Gel and Column techniques
Reagent + Diluent Kit

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service