Skip to Content
Merck
All Photos(1)

Documents

EZHIASF

Millipore

Human Insulin ELISA Kit

measures and quantify insulin levels in 20 μL serum, plasma or cell culture supernatent; animal serum free

Synonym(s):

Insulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

product name

Human Insulin ELISA, animal serum free, This Human Insulin ELISA, animal serum free is used to measure & quantify Insulin levels in Metabolism & Endocrine research.

Quality Level

species reactivity

human

packaging

kit of 1 × 96 wells

parameter

20 μL sample volume (4hr assay)

assay range

accuracy: 88-128%
linearity: 73-100%
sensitivity: 0.73 μU/mL
standard curve range: 2-200 μU/mL

technique(s)

ELISA: suitable

input

sample type cell culture supernatant
sample type serum
sample type plasma (K2 EDTA)

NCBI accession no.

UniProt accession no.

application(s)

research use

detection method

colorimetric (450nm)

shipped in

wet ice

storage temp.

2-8°C

General description

This unique Human Insulin Elisa kit is animal serum free–the monoclonal antibodies are harvested from clones grown in culture medium devoid of any animal protein.

Specificity

No Cross-reactivity with Human Pro-insulin or Human C-peptide

Application

3.5 hour assay, room temperature, 20 µL sample volume
Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders
Used to detect/quantify: Insulin

Disclaimer

For research use only. Not for use in diagnostic procedures.

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service