Skip to Content
Merck
All Photos(5)

Key Documents

EZFITMHTA1

Millipore

EZ-Fit® Manifold Filtration Head

for use with EZ-Fit® Filtration Manifold, pkg of 1 unit

Synonym(s):

EZ-FIT® Manifold Head High Throughput, For bioburden application, filtration head for manifold

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

316L stainless steel

Quality Level

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 1 unit

manufacturer/tradename

EZ-Fit®

application(s)

bioburden testing
bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The EZ-Fit® Manifold Head High Throughput Application has been designed to improve ease of use, reduce maintenance and limit the risk of contamination. These filtration heads offer full compatibility with the EZ-Fit® Filtration Units.

Application

The EZ-Fit® Manifold Head High Throughput Application is an accessory of the EZ-Fit® Manifold used during filtration of the test samples for general microbiological analysis, bioburden testing, and water monitoring.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service