Skip to Content
Merck
All Photos(1)

Key Documents

EZFITL0HE3

Millipore

Microfil® V or Microfil® S Filtration Head

for use with EZ-Fit® Filtration Manifold, availabe as 3 units, pkg of 3 units

Synonym(s):

For use with EZ-Fit® vacuum manifold, Microfil® V or Microfil® S filtration head for EZ-Fit® Manifold (3 units)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

316L stainless steel

Quality Level

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 3 units

manufacturer/tradename

EZ-Fit®

application(s)

bioburden testing
environmental
food and beverages
water monitoring

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service