Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
form
liquid
manufacturer/tradename
Specialty Media, EmbryoMax®
technique(s)
cell culture | stem cell: suitable
input
sample type: mouse embryonic stem cell(s)
sample type induced pluripotent stem cell(s)
Preparation Note
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description The EmbryoMax Filtered Light Mineral Oil is available in a 100 mL format and may be used for routine mouse embryonic stem cell culture applications. | description 5.0% (50′000 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified) | description 4.0% (40′000 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified) | description 0.001% (10 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified) |
| input sample type: mouse embryonic stem cell(s) | input - | input - | input - |
| manufacturer/tradename Specialty Media, EmbryoMax® | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd |
| technique(s) cell culture | stem cell: suitable | technique(s) - | technique(s) - | technique(s) - |
| Quality Level 100 | Quality Level - | Quality Level - | Quality Level - |
| form liquid | form - | form - | form - |
Storage Class
10 - Combustible liquids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

