EFHAB100M3
EZ-Fit® filtration unit with absorbent pad
0.45 µm MCE black membrane with 100 mL funnel, multi packaging of 3 units, suitable for solid and liquid cultivation
Synonym(s):
EZ-Fit® Blue Funnel HABG 100ml multi-pack, EZ-Fit®Filtration funnel, Membrane filtration device made of gridded, mixed cellulose esters membrane for bioburden testing
About This Item
Recommended Products
material
black filter
blue acrylonitrile-butadiene-styrene (ABS) base
cellulose pad(s)
gridded black mixed cellulose esters (MCE) membrane
low-density polyethylene plug
polystyrene cover
Quality Level
Agency
EP
ISO
USP
sterility
sterile; electron beam irradiated
feature
ready-to-use
manufacturer/tradename
EZ-Fit®
packaging
pkg of Multi packaging of 3 units
parameter
100 mL sample volume
capacity
100 mL
filter diam.
47 mm
funnel capacity
100 mL
pore size
0.45 μm pore size
input
sample type food(s)
sample type pharmaceutical(s)
sample type beverage(s)
application(s)
bioburden testing (for liquid samples)
cosmetics
industrial qc
water monitoring
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The filtration units are e-beam irrated, a vented lid covers the sample throughout filtration, and a 360° rim prevents forceps from accidentally touching the membrane filtration area during membrane transfer, to protect samples from contamination while ensuring reliable microbial enumeration results.
The EZ-FIT® Azure feature makes the EZ-FIT® filtration unit even more ergonomic and convenient with transparent funnel and all-around filling level indicators for an easy filling from any angle. The innovative funnel assembly design prevents any leaks or sample by-pass of the membrane.
The blue units provide the extra flexibility of either transferring the membrane to an agar plate or converting the device into a Petri dish by adding the liquid media, after filtration.
The filtration units conform to international standards and minimize sample residue. The units can be stacked to save the work area. This disposable device is designed to fit onto the EZ-FIT® Manifold, which features specific EZ-FIT® Filtration heads.
Application
Features and Benefits
- The transparent funnel allows you to see when filtration is complete from any angle
- Thanks to all-around filling level indicators there is no need to turn the funnel when placing it on the filtration support
- The shape of the membrane base makes transferring the membrane with the forceps easy
- The unit’s design allows the funnel to be removed with one hand
- The units are stackable, saving space in the work area
- It comes with an absorbent pad (integrated into the device)
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology.
Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology.
Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology.
Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service