Skip to Content
Merck

ECPRIME

mPAGE® Primary Electrode Core

replacement part for mPAGE® Mini Gel Tank

Synonym(s):

electrode assembly

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NC.01
UNSPSC Code:
41105305
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


AC/DC input

100 watts (maximum power limit), 300 V (maximum voltage limit)

packaging

pkg of 1 packs

manufacturer/tradename

Millipore

parameter

70 °C ambient temp. limit

technique(s)

SDS-PAGE: suitable, electrophoresis: suitable

compatibility

for use with mPAGE® Mini Gel Tank.

General description

mPAGE® Primary Electrode Core, for use with mPAGE® Mini Gel Tank (MGT-2 and MGT-4).
The mPAGE® Mini Gel Tank is used to perform high-resolution protein separation on handcast or precast polyacrylamide gels. The gel tank is designed for use with 8 cm x 10 cm mini gels and is compatible with gels cast on the mPAGE® Gel Caster and mPAGE® Precast Gels. The mPAGE® Mini Gel Tank is available in 2 formats for running up to 2 or 4 mini gels and is designed to prevent incorrect assembly and leakage.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library