Skip to Content
Merck

DMIDI

DirectLoad Midi Horizontal Electrophoresis System, with 15x7, 10 &15cm gel trays

for agarose gel DNA and RNA horizontal electrophoresis

Synonym(s):

midi horizontal electrophoresis system, midi electrophoresis system, horizontal electrophoresis system, electrophoresis system

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41105340
NACRES:
NA.31
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

For use with DirectLoad™ and DirectLoad™ Midi branded products

Quality Segment

packaging

pkg of 1 unit

technique(s)

electrophoresis: suitable

application(s)

pharmaceutical

General description

The DirectLoad Midi Horizontal Electrophoresis Systems provide researchers flexible and easy-to-use platforms for agarose gel DNA and RNA electrophoresis experiments. Compatible Midi gel trays, combs, and volumes can be modified to research needs. Midi Electrophoresis System products are optimal for large throughput experiments. Product accessory names are matched to ensure compatibility with electrophoresis systems

Application

DirectLoad Midi Horizontal Electrophoresis System, with 15x7, 10 & 15cm gel trays is suitable for medium throughput experiments up to 210 samples depending on configuration.

Features and Benefits

  • Easy-click and color-coded attachments are provided for safety and single-orientation assembly
  • For safety the DirectLoad lid disconnects the power source upon removal
  • DirectLoad Gel Trays and Gel Dams are leak-proof and enable agarose gel preparation in parallel
  • Maximize experimental versatility with multiple gel trays and comb sizes
  • DirectLoad Combs are height adjustable and act as sample loading guides using single and multi-channel pipettes
  • To increase the longevity of the system replacement accessories are available

Other Notes

Midi System Contents Include:
(1) DirectLoad Midi Tank (Including Electrodes) - DMS15-TANK,
(1) DirectLoad Midi Lid - DMS15-LID,
(1) DirectLoad Midi 15 x 7cm Gel Tray - DMS15-TR7,
(1) DirectLoad Midi 15 x 10cm Gel Tray - DMS15-TR10,
(1) DirectLoad Midi 15 x 15cm Gel Tray - DMS15-TR15,
(2 total) DirectLoad Midi Gel Tray Dams - DMS15-DAMS,
(2) DirectLoad Midi comb, 20 sample, 1mm - DMS15-20-1,
(1) DirectLoad Midi Loading Guides - DMS15-LG,
(1) DirectLoad Electrophoresis cables (Black & Red) - DMS-CABLES.

Legal Information

DirectLoad is a trademark of Sigma-Aldrich Co. LLC




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library