Skip to Content
Merck

Skip To

CC10

GM6001 MMP Inhibitor

The GM6001 MMP Inhibitor controls the biological activity of MMP. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

GM6001 Inhibitor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352200
NACRES:
NA.41
eCl@ss:
32160405
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

liquid

manufacturer/tradename

Chemicon®

concentration

1 mg/mL in DMSO, 2.5 mM in DMSO

NCBI accession no.

UniProt accession no.

General description

Specificity: GM6001 MMP inhibitor, also known as Ilomastat or N-[(2R)-2-(hydroxamidocarbonylmethyl)-4-methylpentanoyl]-L-tryptophan methylamide, is a potent inhibitor of collagenases. Molecular weight: 388.47

Reported Ki values are as follows (see Galardy Ref):·

Human MMP-1 (Fibroblast collagenase): 0.4 nM·

Human MMP-3 (Stromelysin): 27 nM·

Human MMP-2 (72 kDa gelatinase): 0.5 nM·

Human MMP-8 (Neutrophil collagenase): 0.1 nM·

Human MMP-9 (92 kDa gelatinase): 0.2 nM

Typical working dilution for use in cultured cells is 10-25 μM. GM6001 will precipitate from aqueous solutions exceeding 100μM. Aqueous dilutions should be performed using low salt buffers to prevent precipitation of GM6001 from solution (Recommended dilution buffer: 50 mM Tris pH7.5, 150 mM NaCl, and 20 mM CaCl2.2H20). It is recommended that the concentration of GM6001 in aqueous working solutions is determined upon dilution and confirmed prior to each use. Additional Application information:
Use in vivo
Typical dosage for GM6001 in animals is 50-100mg/kg body weight. It is generally administered IP every day or every other day. It is thought to cross the blood brain barrier since studies have shown it effectiveness in MS models.

If delivered by I.V. the half life in an animal is approximate an hour so it will have to be administered quite often. Thus the preferred way is to deliver the drug by I.P as a suspension in water or saline. The half life does not change but because it is dissolving in the animal′s system slowly, administration is not done as frequently. It has to be redelivered every other day or so. It can also be delivered subcutaneously in a saline suspension. The drug will precipitate as stay as a mass within the tissue, but surprisingly enough does get into the animal system to be effective.

Solubility
Soluble up to at least 400mg/ml in DMSO. It can then be diluted in water and must be mixed very quickly to avoid precipitation.

Stability
GM6001 in DMSO or Water at a concentration of 1mM (0.1mM is preferred) decomposes at the rate of 1% per month at 4%. At 37C this increases to 1% per day. At -20 it is quite stable and as a solid very highly stable
Low salt buffers are recommended as diluents to reduce precipitation. Diluted solutions should never be frozen.

Concentration of the stock solution should be periodically determined by absorbance at 280nm.

Biochem/physiol Actions

Inhibitor Type: Protease Inhibitors
Protein Target: Interstitial collagenase

Physical form

Provided at 1 mg/mL (2.5 mM) in DMSO.

Preparation Note

Maintain frozen at -20°C in DMSO for up to 12 months. Avoid storage once diluted into aqueous solutions.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CC1100364205444225
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

liquid

form

powder

form

solid

form

solid

UniProt accession no.

P03956, P08254, P22894, P08253

UniProt accession no.

P03956, P08253, P08254, P22894

UniProt accession no.

-

UniProt accession no.

-

NCBI accession no.

NM_002421.2

NCBI accession no.

NM_002421.2

NCBI accession no.

-

NCBI accession no.

-

manufacturer/tradename

Chemicon®

manufacturer/tradename

Chemicon®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

concentration

1 mg/mL in DMSO

concentration

-

concentration

-

concentration

-


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

188.6 °F - (refers to pure substance)

flash_point_c

87 °C - (refers to pure substance)



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ji Yoon Park et al.
Journal of biochemistry, 151(3), 291-298 (2011-12-16)
Screening of matrix metalloproteinase (MMP)-14 substrates in human plasma using a proteomics approach previously identified apolipoprotein A-IV (apoA-IV) as a novel substrate for MMP-14. Here, we show that among the tested MMPs, purified apoA-IV is most susceptible to cleavage by
Low molecular weight inhibitors in corneal ulceration.
R E Galardy et al.
Annals of the New York Academy of Sciences, 732, 315-323 (1994-09-06)
Matthias P Lutolf et al.
Nature biotechnology, 21(5), 513-518 (2003-04-22)
We have engineered synthetic poly(ethylene glycol) (PEG)-based hydrogels as cell-ingrowth matrices for in situ bone regeneration. These networks contain a combination of pendant oligopeptide ligands for cell adhesion (RGDSP) and substrates for matrix metalloproteinase (MMP) as linkers between PEG chains.



Global Trade Item Number

SKUGTIN
CC100004053252336256
CC101004053252731969

Questions

Reviews

No rating value

Active Filters