Skip to Content

CC076

Human Collagen Type IV

from human placenta, liquid, 1 mg/mL, suitable for cell culture, used for gel formation

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75
Biological source:
human
Form:
liquid, solution
Technique(s):
cell culture | mammalian: suitable
Concentration:
1 mg/mL
Mol wt:
300 kDa
Assay:
95% (Human collagen type IV, SDS-PAGE)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Human Collagen Type IV, 1 mg/mL, solution, BioReagent, suitable for cell culture

biological source

human

Quality Segment

assay

95% (Human collagen type IV, SDS-PAGE)

form

liquid, solution

mol wt

300 kDa

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

cell culture | mammalian: suitable

impurities

<5% Human collagen types I-III, V, VI, and non-collagen proteins

input

sample type epithelial cells
sample type hematopoietic stem cell(s)
sample type pancreatic stem cell(s)
sample type neural stem cell(s)
sample type mesenchymal stem cell(s)
sample type induced pluripotent stem cell(s)
sample type: human embryonic stem cell(s)

solubility

water: soluble at 20 °C

NCBI accession no.

UniProt accession no.

binding specificity

Peptide Source: Laminin
, Peptide Source: Nidogen

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... COL4A1(1282)

General description

Human collagen type IV is a major structural element predominantly found in the basement membrane of the skin. It is a native triple helix and heterotrimeric protein containing two α1-like and one α2-like genes. Type 4 collagen forms a sheet-like extracellular matrix in multicellular tissues that exists around certain cell types like adipocytes, skeletal muscle cells, heart muscle cells, and smooth muscle cells. Human Collagen IV is extracted from washed dissected tissue into dilute acetic acid after mild pepsin treatment. The Collagen type 4 was purified by using differential salt precipitation. Purity and retention of the native helical structure were controlled by SDS-PAGE and by reaction with anti-collagen type-specific monoclonal antibodies. Molecular weight: 300kDa; Solubility: 10mg/mL Product Source: Human placenta, negative for HBsAg and HIV antibodies

Application

Collagen IV from the human placenta is suitable for use as a coating material for cell culture studies and the formation of collagen gels. Human Collagen Type IV has been used: to prepare collagen gel for the invasion assay to provide a supportive matrix for assessing the invasive capacity of monocytes in a cell culture systemas an extracellular matrix (ECM) protein in microarray analysis, to coat nitrocellulose film slidesto coat the microchannels of the P96-O2 plate, and provide a surface for the adhesion and growth of endothelial cells (hMVECs) and renal proximal tubular epithelial cells (hRPTECs) in a co-culture system

Biochem/physiol Actions

Human collagen type IV is associated with Alport and Goodpasture′s syndrome, as well as several rheumatological and dermatological diseases including, acquired epidermolysis bullosa.

Physical form

Liquid purified protein. In 100 μL of 0.5M acetic acid solution, pH 2.5. Salt-free, no preservative.

Preparation Note

Store at -20ºC for up to 12 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Retention of native structure was confirmed by ability to form microfibrils.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library