Skip to Content
Merck
All Photos(1)

Documents

CBL171-I

Sigma-Aldrich

Anti-Actin, smooth muscle Antibody, clone ASM-1/1A4

clone ASM-1 (1A4), from mouse

Synonym(s):

Actin, aortic smooth muscle, Alpha-actin-2, Cell growth-inhibiting gene 46 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ASM-1 (1A4), monoclonal

species reactivity

mouse, chicken, rat, human, bovine

species reactivity (predicted by homology)

equine (based on 100% sequence homology)

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACTA2(59)

General description

Actin, aortic smooth muscle (UniProt P62736; also known as Alpha-actin-2, Cell growth-inhibiting gene 46 protein) is encoded by the ACTA2 (also known as AAT6, ACTSA, ACTVS, GIG46, MYMY5) gene (Gene ID 59) in human. Actins are globular multi-functional proteins that serve as the basic building blocks of cytoskeletal microfilaments and are among the most conserved eukaryotic proteins. Six actin types exist, skeletal muscle alpha-actin is encoded by the ACTA1 gene, smooth muscle alpha-actin by the ACTA2 gene, cytoplasmic beta-actin by the ACTB gene, cardiac muscle alpha-actin by the ACTC1 gene, cytoplasmic gamma-actin by the ACTG1 gene, and smooth muscle gamma-actin by the ACTG2 (a.k.a. ACTA3) gene. Although actins show >90% overall sequence homology, isoforms do show spatial, temporal, and tissue-specific expresson patterns and only 50-60% homology is found in their 18 N-terminal residues. Cytoplasmic β and γ-actins, are thought to be present in all cells, while the other four actin isoforms are typically found in specific adult muscle tissue types. Actins exist in a variety of structural states, depending on the specific ionic conditions or the interaction with ligand proteins. The oligomeric and polymeric forms that actin molecules assume are dependent on the distinct conformations they adopt.

Specificity

Clone ASM-1, also known as anti-αsm-1 and clone 1A4, reacted with aortic actin, but not actin from fibroblasts (β- and γ-cytoplasmic), striated muscle (α-sarcomeric), or myocardium (α-myocardial) by ELISA and Western blotting (Skalli, O., et al. (1986). J. Cell Biol. 103(6 Pt 2):2787-2796).

Immunogen

BSA-conjugated linear peptide corresponding to the N-terminal sequence of human smooth muscle actin.
Epitope: N-terminus.

Application

Anti-Actin, smooth muscle Antibody, clone ASM-1/1A4 is an antibody against Actin for use in Western Blotting, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, ELISA.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling
Western Blotting Analysis: A representative lot detected full-length and caspse-3-cleaved smooth muscle actin in uterus extracts from pregnant mice (Jeyasuria, P., et al. (2009). Biol. Reprod. 80(5):928-934).
Western Blotting Analysis: A representative lot detected smooth muscle actin in PC-3M-LN4 and PC-3M-LN4-derived human prostate cancer cells (Zvieriev, V., et al. (2005). Biochem. Biophys. Commun. 337(2):498-504).
Western Blotting Analysis: A representative lot detected developing stage-dependent vascular alpha-actin level in chicken heart of various somite stages (Ehler, E., et al. (2004). Dev. Dyn. 229(4):745-755).
Western Blotting Analysis: A representative lot detected aortic actin, but not actin from fibroblasts (beta- and gamma-cytoplasmic), striated muscle (alpha-sarcomeric), or myocardium (alpha-myocardial) in various human, rat, bovine and rabbit tissue and cell lysates (Skalli, O., et al. (1986). J. Cell Biol. 103(6 Pt 2):2787-2796).
Immunohistochemistry Analysis: A representative lot immunostained smooth muscle actin-positive myofibroblasts in paraformaldehyde-fix rat skin wounds sections (Mirastschijski, U., et al. (2010). Wound Repair Regen. 18(2):223-234).
Immunohistochemistry Analysis: A representative lot immunostained activated myofibroblasts in formalin-fixed, paraffin-embedded mouse submandibular salivary gland (Hall, B.E., et al. (2010). Lab. Invest. 90(4):543-555).
Immunohistochemistry Analysis: A representative lot immunostained α-SMA-positive cells in paraffin-embedded rat healing patellar tendon sections (Fu, S.C., et al. (2008). J. Orthop. Res. 26(3):374-383).
Immunocytochemistry Analysis: Representative lots immunostained the mesoderm layer of embryoid bodies formed in vitro from northern white rhinoceros iPSCs, as well as murine iPSCs and mESCs (Ben-Nun, I.F., et al. (2011). Nat. Methods. 8(10):829-831; Gerlach, J.C., et al. (2010). Cells Tissues Organs. 192(1):39-49; Shao, L., et al. (2009). Cell Res. 19(3):296-306).
Immunohistochemistry Analysis: A representative lot immunostained a subsets of cultured rat aortic medial smooth muscle cells (SMCs) and dermal fibroblasts (Skalli, O., et al. (1986). J. Cell Biol. 103(6 Pt 2):2787-2796).
Immunohistochemistry Analysis: A representative lot immunostained cultured rat tendon cells (Fu, S.C., et al. (2008). J. Orthop. Res. 26(3):374-383).
Immunofluorescence Analysis: A representative lot immunostained myofibrils using methanol-fixed, 9-somite stage chicken heart whole-mount preparations. The vascular alpha-actin staining started to disappear from a subset of the myofibrils when using 10-somite stage chicken heart preparations (Ehler, E., et al. (2004). Dev. Dyn. 229(4):745-755).
Immunofluorescence Analysis: A representative lot immunostained stromal vessel SM cells in acetone-fixed benign (leiomyomas) and malignant (leiomyosarcomas & intravascular leiomyomatosis) smooth muscle (SM) neoplasms cryosections (Schürch, W., et al. (1987). Am. J. Pathol. 128(1):91-103).
Immunofluorescence Analysis: A representative lot immunostained chicken gizzard and rat myocardium blood vessels, as well as smooth muscle cells (SMCs) in various acetone-fixed human and rat cryosections, whereas chicken gizzard parenchyrnal cells, rat cardiocytes, aorta endothelial cells and adventitial fibroblasts were negative (Skalli, O., et al. (1986). J. Cell Biol. 103(6 Pt 2):2787-2796).
ELISA Analysis: A representative lot detected aortic actin, but not actin from fibroblasts (beta- and gamma-cytoplasmic), striated muscle (alpha-sarcomeric), or myocardium (alpha-myocardial) by ELISA (Skalli, O., et al. (1986). J. Cell Biol. 103(6 Pt 2):2787-2796).

Quality

Evaluated by Western Blotting in HUVEC lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected smooth muscle actin in 10 µg of HUVEC lysate.

Target description

~45 kDa observed.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tzu-Pin Shentu et al.
Scientific reports, 7(1), 18052-18052 (2017-12-24)
Bone marrow-derived mesenchymal stem cells (MSC) have been promoted for multiple therapeutic applications. Many beneficial effects of MSCs are paracrine, dependent on extracellular vesicles (EVs). Although MSC-derived EVs (mEVs) are beneficial for acute lung injury and pulmonary fibrosis, mechanisms of
Kazutoshi Takahashi et al.
PLoS genetics, 17(5), e1009587-e1009587 (2021-05-26)
Human pluripotent stem cells (PSCs) express human endogenous retrovirus type-H (HERV-H), which exists as more than a thousand copies on the human genome and frequently produces chimeric transcripts as long-non-coding RNAs (lncRNAs) fused with downstream neighbor genes. Previous studies showed
Clara Dees et al.
The Journal of clinical investigation, 130(5), 2347-2363 (2020-01-29)
Fibroblasts are key effector cells in tissue remodeling. They remain persistently activated in fibrotic diseases, resulting in progressive deposition of extracellular matrix. Although fibroblast activation may be initiated by external factors, prolonged activation can induce an "autonomous," self-maintaining profibrotic phenotype
Indrajit Nanda et al.
Cells, 11(18) (2022-09-24)
Hepatic stellate cells (HSCs) are also known as lipocytes, fat-storing cells, perisinusoidal cells, or Ito cells. These liver-specific mesenchymal cells represent about 5% to 8% of all liver cells, playing a key role in maintaining the microenvironment of the hepatic
Kiara Gäberlein et al.
Cells, 12(12) (2023-06-28)
The rat hepatic stellate cell line PAV-1 was established two decades ago and proposed as a cellular model to study aspects of hepatic retinoic acid metabolism. This cell line exhibits a myofibroblast-like phenotype but also has the ability to store

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service