Skip to Content
Merck

Skip To

CBA059

Annexin V-FITC Apoptosis Detection Kit II

fluorometric assay kit for measuring Annexin V in apoptotic cells.

Synonym(s):

Annexin V Assay, Apoptosis Detection Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.84
UNSPSC Code:
41116158
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

usage

sufficient for 20 tests

packaging

pkg of 1 kit

manufacturer/tradename

Calbiochem®

technique(s)

flow cytometry: suitable, immunofluorescence: suitable

detection method

fluorometric

shipped in

wet ice

storage temp.

2-8°C

General description

A convenient kit for the identification of changes in the plasma membrane that occur during apoptosis. In apoptotic cells, phosphoatidylserine (PS) is translocated from the inner to the outer leaflet of the plasma membrane, allowing for the detection of PS on the cell surface. In the presence of Ca 2+, Annexin V exhibits a high affinity for PS and binds cells with exposed PS. Annexin V, when conjugated to a fluorophore, can be used to monitor apoptopic cells using flow cytometry or immunofluorescence microscopy.

Other Notes

Kit contains:
  • Annexin V-FITC
  • Binding Buffer
  • Propidium Iodide
  • user protocol.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
APOAFQIA113S7110
technique(s)

flow cytometry: suitable, immunofluorescence: suitable

technique(s)

flow cytometry: suitable

technique(s)

flow cytometry: suitable

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

ApopTag, Chemicon®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

-

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

dry ice

usage

sufficient for 20 tests

usage

(20 tests)

usage

sufficient for 100 tests

usage

-

detection method

fluorometric

detection method

fluorometric

detection method

fluorometric

detection method

fluorometric


Storage Class

10 - Combustible liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters