BPCKHF18002S25
Hi-Flow™ Plus Membrane 180
2mil backing, 25mm width, pack of 11 slit rolls
Synonym(s):
Hi-Flow, HF, lateral flow membrane, Immunoassay membranes
About This Item
Quality Level
packaging
pack of 11 roll
manufacturer/tradename
Hi-Flow Plus
parameter
180 sec/4 cm capillary flow rate
backing thickness
2 mil (50 μm)
filter W × L
25 mm × 100 m
roll L
100 m (328 ft)
slit W
25 mm
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Consistent Flow & Surface Chemistry, Robust manufacturing process
- Proprietary Surface Chemistry Optimised for Lateral Flow Applications
- High Quality Level: MQ400
- Trusted Partner and Brand with Decades of membrane manufacturing experience
- High production capacity, high stock levels, multiple manufacturing lines
- Convenient samples packs available in several formats
- Manufacturing scale quantities available to purchase online with immediate shipping.
BENEFITS:
- Consistent capillary flow time and membrane thickness
- Superior surface morphology allows for consistent and even flow distribution of reagents across the membrane surface
- Sharp, intense, and uniform test and control line formation
- Excellent intra-lot and inter-lot consistency.
- Better Assay sensitivity
- Excellent reproducibility
- Improved lateral flow test manufacturing yields.
- Quick Delivery and Lead time.
- No supply disruption
- Excellent Technical support
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service