AP307A
Goat Anti-Rabbit IgG Antibody, (H+L) Alkaline Phosphatase conjugate
1.0 mg/mL, Chemicon®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46
Recommended Products
biological source
goat
Quality Level
conjugate
alkaline phosphatase conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
rabbit
manufacturer/tradename
Chemicon®
concentration
1.0 mg/mL
technique(s)
ELISA: suitable
western blot: suitable
shipped in
wet ice
target post-translational modification
unmodified
Specificity
Specific for rabbit IgG, heavy and light chain. The cross-reactivities of anti-rabbit IgG antibody are tested in an ELISA. Cross-reactivity to human and mouse IgG is <2% of the reactivity to the homologous antigen.
Application
Detect Rabbit IgG using this Goat anti-Rabbit IgG Antibody, (H+L) Alkaline Phosphatase conjugate validated for use in ELISA & WB.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M L Katz et al.
Journal of inherited metabolic disease, 30(6), 952-963 (2007-11-16)
The neuronal ceroid lipofuscinoses (NCLs) are inherited neurodegenerative diseases characterized by massive accumulation of autofluorescent storage bodies in neurons and other cells. A late-onset form of NCL occurs in Tibetan terrier dogs. Gel electrophoretic analyses of isolated storage body proteins
Zibin Tan et al.
Nature communications, 15(1), 3420-3420 (2024-04-25)
Poly-β-(1-6)-N-acetylglucosamine (PNAG) is an important vaccine target, expressed on many pathogens. A critical hurdle in developing PNAG based vaccine is that the impacts of the number and the position of free amine vs N-acetylation on its antigenicity are not well
Edita Holásková et al.
Biotechnology journal, 13(6), e1700628-e1700628 (2018-01-26)
The peptide LL-37, a component of the human innate immune system, represents a promising drug candidate. In particular, the development of low-cost production platform technology is a critical bottleneck in its use in medicine. In the present study, a viable
Liang-Chen Su et al.
Frontiers in plant science, 6, 512-512 (2015-07-29)
Histone acetylation, which together with histone methylation regulates gene activity in response to stress, is an important epigenetic modification. There is an increasing research focus on histone acetylation in crops, but there is no information to date in peanut (Arachis
Francesco De Rose et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 60(6), 770-776 (2018-10-27)
Preoperative characterization of thyroid nodules is challenging since thyroid scintigraphy fails to distinguish between benign and malignant lesions. Galectin-3 (gal-3) is expressed in well-differentiated and in undifferentiated thyroid cancer types but not in normal thyrocytes and benign thyroid lesions. Herein
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service