Skip to Content
Merck
All Photos(1)

Key Documents

AP183C

Sigma-Aldrich

Goat Anti-Rat IgG Antibody, Cy3 conjugate, Species Adsorbed

1.4 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

CY3 conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

concentration

1.4 mg/mL

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Reacts with heavy chain of Rat IgG, as well as the light chains from most rat immunoglobulin classes.

Monospecific on IEP against whole Mouse Serum.

FLOUROPHORE/PROTEIN RATIO:

A552/A280 = 2.5

Immunogen

Highly purified Rat IgG isolated from pooled normal Mouse Serum.

Application

1:100 - 1:800 for most applications.
Goat anti-Rat IgG Antibody, Cy3 conjugate, Species Adsorbed is an antibody against Rat IgG for use in IF.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling

Storage and Stability

Store at +2-8°C. Do not freeze. After opening, restore to 0.4mL with distilled water. As an undiluted liquid the product is stable at 2-8°C for several weeks. For extended storage after reconstitution, we suggest the addition of an equal volume of glycerol to make a final glycerol concentration of 50% followed by storage at -20°C, with or without aliquoting. Exp. Date: 1 year from reconstitution.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sohei Suzuki et al.
Development, growth & differentiation, 63(7), 343-353 (2021-09-05)
Germ layer formation is driven by embryonic cell sorting during the early developmental stages. Starfish (Patiria pectinifera) embryos have a connected endoderm and ectoderm, albeit with few contact surfaces between the epithelia. To better understand the association between cell sorting
Marie-Therese Kurzbauer et al.
The Plant cell, 30(2), 415-428 (2018-01-21)
Fanconi anemia (FA) is a human autosomal recessive disorder characterized by chromosomal instability, developmental pathologies, predisposition to cancer, and reduced fertility. So far, 19 genes have been implicated in FA, most of them involved in DNA repair. Some are conserved
Masato Uemura et al.
Scientific reports, 12(1), 11933-11933 (2022-07-14)
Protocadherin 9 (Pcdh9) is a member of the cadherin superfamily and is uniquely expressed in the vestibular and limbic systems; however, its physiological role remains unclear. Here, we studied the expression of Pcdh9 in the limbic system and phenotypes of
The role of T-cadherin in axonal pathway formation in neocortical circuits.
Hayano, Y; Zhao, H; Kobayashi, H; Takeuchi, K; Norioka, S; Yamamoto, N
Development null
Vanessa Kurth et al.
The Journal of biological chemistry, 299(8), 104997-104997 (2023-07-03)
Presenilin-1 (PSEN1) is the catalytic subunit of the intramembrane protease γ-secretase and undergoes endoproteolysis during its maturation. Heterozygous mutations in the PSEN1 gene cause early-onset familial Alzheimer's disease (eFAD) and increase the proportion of longer aggregation-prone amyloid-β peptides (Aβ42 and/or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service