Skip to Content
Merck
All Photos(1)

Key Documents

AP180

Sigma-Aldrich

Donkey Anti-Goat IgG Antibody, Species Adsorbed

~1 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

donkey

Quality Level

conjugate

unconjugated

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

goat

manufacturer/tradename

Chemicon®

concentration

~1 mg/mL

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Goat IgG (H+L) (absorbed for minimal cross-reactivity to Chicken, Guinea Pig, Syrian Hamster, Horse, Human, Mouse, Rabbit, and Rat Serum Proteins)

Application

Donkey anti-Goat IgG Antibody, Species Adsorbed detects level of Donkey Goat IgG & has been published & validated for use in ELISA, IP & WB.
ELISA and Western Blotting at 1:1000-1:10,000.Optimal working dilution must be determined by the end user. WARNING: Dilute this antibody with PBS or TBS only. Use of bovine serum or dry milk may contain IgG which reacts with various IgG antibodies. Use of BSA or Milk can cause background and reduce secondary antibody titer.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling

Physical form

Liquid in 0.1M Sodium Phosphate, 0.25M NaCl, pH 7.6

Storage and Stability

Store product at 2-8′C under sterile conditions for up to 12 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Kamiguchi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 18(14), 5311-5321 (1998-07-03)
Cell-cell interactions mediated via cell adhesion molecules (CAMs) are dynamically regulated during nervous system development. One mechanism to control the amount of cell surface CAMs is to regulate their recycling from the plasma membrane. The L1 subfamily of CAMs has
Xu Zeng et al.
Botanical studies, 58(1), 31-31 (2017-07-26)
Gastrodia elata is a well-known medicinal orchid. In nature, the germination rate of G. elata is extremely poor, because there is no endosperm within the mature seed. It is crucial for G. elata to obtain nutrition from mycorrhizal fungi (Mycena)
R Jiang et al.
The Journal of biological chemistry, 275(12), 8439-8447 (2000-03-18)
When Hsc70 uncoats clathrin-coated vesicles in an auxilin- and ATP-dependent reaction, a single round of rapid uncoating occurs followed by very slow steady-state uncoating. We now show that this biphasic time course occurs because Hsc70 sequentially forms two types of
Narcoleptic orexin receptor knockout mice express enhanced cholinergic properties in laterodorsal tegmental neurons.
Kalogiannis, M; Grupke, SL; Potter, PE; Edwards, JG; Chemelli, RM; Kisanuki et al.
The European Journal of Neuroscience null
Chunjiang Yu et al.
Molecular neurodegeneration, 5, 19-19 (2010-05-19)
One pathological hallmark of Alzheimer's disease (AD) is amyloid plaques, composed primarily of amyloid-beta peptide (Abeta). Over-production or diminished clearance of the 42 amino acid form of Abeta (Abeta42) in the brain leads to accumulation of soluble Abeta and plaque

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service