Skip to Content
Merck
All Photos(2)

Documents

ABT123

Sigma-Aldrich

Anti-Collagen Type I Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Collagen alpha-1(I) chain, Alpha-1 type I collagen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... COL1A1(1277)

General description

Collagens are highly conserved throughout evolution and are characterised by an uninterrupted "Glycine X Y" triplet repeat that is a necessary part of the triple helical structure. Type I collagen (95 kDa) is found in bone, cornea, skin and tendon. Mutations in the encoding gene are associated with osteogenesis imperfecta, Ehlers Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for Platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor.

Immunogen

Rat tail collagen

Application

Detect Collagen Type I using this Anti-Collagen Type I Antibody validated for use in Western Blotting, IHC(P), ELISA.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
Specificity ELISA Analysis: A representative lot detected mouse and rat collagen but not human collagen in ELISA.

Western Blot Analysis: A 1:2,000 dilution from a representative lot detected Collagen Type 1 on 10 µg of rat skin tissue lysate.

Quality

Evaluated by Immunohistochemistry in rat skin tissue.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected Collagen Type 1 in rat skin tissue.

Target description

130 kDa calculated

Linkage

Replaces: AB755P

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat skin tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lihua Cui et al.
Frontiers in pharmacology, 12, 679557-679557 (2021-06-29)
Pancreatic fibrosis is a pathological characteristic of chronic pancreatitis (CP) and pancreatic cancer. Chaihu Guizhi Ganjiang Decoction (CGGD) is a traditional Chinese medicine, which is widely used in the clinical treatment of digestive diseases. However, the potential anti-fibrosis mechanism of
Takashi Ohnishi et al.
Cell death & disease, 14(7), 447-447 (2023-07-20)
Pathological mineralization of intervertebral disc is debilitating and painful and linked to disc degeneration in a subset of human patients. An adenosine triphosphate efflux transporter, progressive ankylosis (ANK) is a regulator of extracellular inorganic pyrophosphate levels and plays an important
Yitong Zhang et al.
Molecular medicine reports, 19(1), 115-124 (2018-11-16)
Numerous studies have revealed that microRNAs (miRNAs) are functional non‑coding RNAs that serve roles in a variety of biological processes. However, the expression patterns and regulatory networks, as well as the miRNAs involved in liver fibrosis remain to be elucidated.
Zilong Xiao et al.
Journal of cardiovascular pharmacology, 82(2), 104-116 (2023-05-10)
Studies have demonstrated the roles of trimetazidine beyond being an antianginal agent in ischemic heart disease (IHD) treatment associated with mechanisms of calcium regulation. Our recent studies revealed that mitochondrial calcium uniporter (MCU, the pore-forming unit responsible for mitochondrial calcium
Shira N Johnston et al.
Frontiers in cell and developmental biology, 12, 1360376-1360376 (2024-03-21)
Hypoxia-inducible factors (HIFs) are essential to the homeostasis of hypoxic tissues. Although HIF-2α, is expressed in nucleus pulposus (NP) cells, consequences of elevated HIF-2 activity on disc health remains unknown. We expressed HIF-2α with proline to alanine substitutions (P405A; P531A)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service