Skip to Content
Merck

ABS45

Anti-phospho-MYPT1 (Thr696) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Myosin phosphatase target subunit 1, Myosin phosphatase-targeting subunit 1, Protein phosphatase myosin-binding subunit, myosin phosphatase, target subunit 1, protein phosphatase 1, regulatory (inhibitor) subunit 12

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
WB, inhibition assay
Citations:
46
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

(~130 kDa observed; 115.28 kDa calculated. Uncharacterized bands may be observed in some lysate(s).)

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

canine (based on 100% sequence homology), rat (based on 100% sequence homology), primate (based on 100% sequence homology), mouse (based on 100% sequence homology), chicken (based on 100% sequence homology), horse (based on 100% sequence homology), Xenopus (based on 100% sequence homology)

technique(s)

inhibition assay: suitable (Peptide Inhibition Assay), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr696)

Gene Information

human ... PPP1R12A(4659)

General description

Anti-phospho-MYPT1 (Thr696), Cat. No. ABS45, is a rabbit polyclonal antibody that detects MYPT1 phosphorylated on Threonine 696 and is tested for use in Western Blotting and Peptide Inhibition Assay.
Protein phosphatase 1 regulatory subunit 12A (UniProt: O14974; also known as Myosin phosphatase-targeting subunit 1, Myosin phosphatase target subunit 1, Protein phosphatase myosin-binding subunit) is encoded by the PPP1R12A (also known as MBS, MYPT1) gene (Gene ID: 4659) in human. Myosin light chain phosphatase with its regulatory subunit, myosin phosphatase target subunit 1 (MYPT1), modulates Ca2+-dependent phosphorylation of myosin light chain by myosin light chain kinase, which is essential for smooth muscle contraction. Myosin phosphatase regulates the interaction of actin and myosin downstream of the guanosine triphosphatase Rho, which inhibits myosin phosphatase through the action of Rho-kinase. MYPT1 is localized on stress fibers and is distributed close to the cell membrane and at cell-cell contacts to regulate myosin phosphatase activity. It is phosphorylated cooperatively by ROCK1 and CDC42BP on Thr-696 that inhibits PP1C activity by an intramolecular mechanism. Mutations in PPP1R12A are involved in Genitourinary and/or brain malformation syndrome (GUBS), an autosomal dominant syndrome that is characterized by multiple congenital anomalies including urogenital malformations and brain abnormalities. Five isoforms of MYPT1 have been described that are produced by alternative splicing. (Ref.: Qiao, Y-N., et al. (2014). J. Biol. Chem. 289(32); 22512-22523).
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide

Immunogen

Epitope: Phosphorylated Thr696
KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphothreonine 696 of human MYPT1.

Application

Anti-phospho-MYPT1 (Thr696) Antibody is an antibody against phospho-MYPT1 (Thr696) for use in WB.
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases
Western Blotting (SNAP ID)Analysis: 0.1 µg/mL from a previous lot detected MYPT1 on 10 µg of NIH/3T3 cell lysate.

Biochem/physiol Actions

This rabbit polyclonal antibody specifically detects MYPT1 phosphorylated on threonine 696.

Physical form

Affinity purified
Purified rabbit polyclonal containing 0.1 M Tris-glycine, pH 7.4, 150 mM NaCl and 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Calyculin/Okadaic treated and untreated HEK293 cell lysates
Evaluated by Western Blotting in HEK293 cell lysate.
Western Blotting Analysis: 0.1 µg/mL of this antibody was used to detect MYPT1 phosphorylated at Thr696 on 10 µg of Calyculin/Okadaic treated and untreated HEK293 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 07-251

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Western blotting is one of the most commonly used techniques in the lab, yet difficulties persist in obtaining consistent, quality results. We’ve been helping scientists publish their Western blots for decades, with continued innovation and steadfast technical support. Explore our expanded portfolio of products, including optimized reagents for chemiluminescent and Ḁuorescent Westerns, as well as the SNAP i.d.® system, which reduces blocking, washing and antibody incubation time from hours to minutes.

Protein Blotting Survival Handbook: Tips and Tricks


Hector N Aguilar et al.
Journal of cellular and molecular medicine, 16(12), 2978-2989 (2012-09-06)
Phosphorylation of myosin regulatory light chain (RLC) triggers contraction in smooth muscle myocytes. Dephosphorylation of phosphorylated RLC (pRLC) is mediated by myosin RLC phosphatase (MLCP), which is negatively regulated by rho-associated kinase (ROK). We have compared basal and stimulated concentrations
Mechanisms underlying potentiation of endothelin-1-induced myofilament Ca(2+) sensitization after subarachnoid hemorrhage.
Kikkawa, Y; Matsuo, S; Kameda, K; Hirano, M; Nakamizo, A; Sasaki, T; Hirano, K
Journal of Cerebral Blood Flow and Metabolism null
Rho-kinase inhibition alleviates pulmonary hypertension in transgenic mice expressing a dominant-negative type II bone morphogenetic protein receptor gene.
Yasuda, T; Tada, Y; Tanabe, N; Tatsumi, K; West, J
American Journal of Physiology. Lung Cellular and Molecular Physiology null



Global Trade Item Number

SKUGTIN
ABS4504053252280634