Skip to Content
Merck
All Photos(2)

Key Documents

ABN241

Sigma-Aldrich

Anti-GluR1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Glutamate receptor 1, GluR-1, AMPA-selective glutamate receptor 1, GluR-A, GluR-K1, Glutamate receptor ionotropic, AMPA 1, GluA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRIA1(2890)
mouse ... Gria1(14799)
rat ... Gria1(50592)

General description

Glutamate receptors (GluRs) are a diverse group responsible for mediating most of the excitatory synaptic transmission in the CNS of vertebrates. They can be categorized as ionotropic or metabotropic and subcategorized by their agonist preferences (NMDA, AMPA or Kainic acid). There are four types of AMPA selective GluR subunits (GluR1, GluR2, GluR3 and GluR4). Tetrameric or pentameric combinations of different subunits contributes to the functional diversity of AMPA receptors. AMPA receptors mediate fast synaptic current at most excitatory synapses, with stoichiometry characterized by subtype composition. The critical residue controlling calcium permeability is in the pore loop region. In GluR1, GluR3, and GluR4, this position is occupied by a Gln residue. The insertion or removal of GluR1/GluR4 oligomeric channels from postsynaptic membranes appears to be LTP/LTD activity dependent while GluR2/GluR3 oligomers are continuously cycling.

Specificity

This antibody recognizes the extracellular domain of GluR1.

Immunogen

KLH-conjugated linear peptide corresponding to the extracellular domain of rat GluR1.

Application

Anti-GluR1 Antibody is a highly specific rabbit polyclonal antibody, that targets GluR1 & has been tested in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected GluR1 in human brain and human cerebellum tissue.

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected GluR1 in 10 µg of rat brain tissue lysate.

Target description

~110 kDa observed

Linkage

Replaces: PC246-100UG

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine J Sellers et al.
Alzheimer's & dementia : the journal of the Alzheimer's Association, 14(3), 306-317 (2017-10-23)
Synapse loss is the structural correlate of the cognitive decline indicative of dementia. In the brains of Alzheimer's disease sufferers, amyloid β (Aβ) peptides aggregate to form senile plaques but as soluble peptides are toxic to synapses. We previously demonstrated
Tao Wu et al.
Journal of biomedical science, 26(1), 79-79 (2019-10-21)
Neuronal activity-induced changes in gene expression patterns are important mediators of neuronal plasticity. Many neuronal genes can be activated or inactivated in response to neuronal depolarization. Mechanisms that activate gene transcription are well established, but activity-dependent mechanisms that silence transcription
Dipen Rajgor et al.
Cell reports, 31(12), 107785-107785 (2020-06-25)
Molecular mechanisms underlying plasticity at brain inhibitory synapses remain poorly characterized. Increased postsynaptic clustering of GABAA receptors (GABAARs) rapidly strengthens inhibition during inhibitory long-term potentiation (iLTP). However, it is unclear how synaptic GABAAR clustering is maintained to sustain iLTP. Here
Shikha Snigdha et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(12), 3611-3622 (2016-03-26)
An increasing number of studies show that an altered epigenetic landscape may cause impairments in regulation of learning and memory-related genes within the aged hippocampus, eventually resulting in cognitive deficits in the aged brain. One such epigenetic repressive mark is
Plinio D Favaro et al.
PLoS biology, 16(12), e2006838-e2006838 (2018-12-27)
The disc-large (DLG)-membrane-associated guanylate kinase (MAGUK) family of proteins forms a central signaling hub of the glutamate receptor complex. Among this family, some proteins regulate developmental maturation of glutamatergic synapses, a process vulnerable to aberrations, which may lead to neurodevelopmental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service